Learn More
GVS Z-link respirator Zytec FR Chin Seal for PX5™ PAPR
The RPB Z-Link is the most versatile multi-purpose respirator on the market. Designed to be resilient in harsh industrial applications while practical to meet the demands of healthcare and life science settings.
Brand:  GVS 16-010-31-CE
Product Code. 18419655
  • 626.00 € / Each
Estimated Shipment: 23-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
The RPB Z-Link is the most versatile multi-purpose respirator on the market. Designed to be resilient in harsh industrial applications while practical to meet the demands of healthcare and life science settings. The Z-Link has been designed with your comfort and functionality in mind, ensuring no matter where you are working, you will be protected for the best moments.
  • Lightweight (880 g) and very comfortable using multi-adjustment helmet system.
  • Superior Comfort: the fully adjustable Comfort-Link fit system allows for total control of your sizing through supportive padding and the adjustable 360 degree head band and ratchet system.
  • Certified to EN 12941 1998 +A2:2008 (TH2 with PX5™ PAPR) and EN 166 2001 1 B T 9 3.
  • Technical Specification EN 397:2012 +A2:2012 (Impact Clauses 5.1.1, 5.1.2, 5.1.3) and
  • Optional Clauses: Very low temperature -300C, molten metal, lateral deformity, electrical properties 440v a.c.
  • Includes: 16-810 Safety Lens, 16-731 Chin Seal Zytec FR, 04-831 PX5™ Breathing Tube
  • To use with PAPR PX5™
Specifications
Specifications
Green
Polycarbonate
One Size
Reusable
Respirator Helmet
No
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
GVS Z-link respirator Zytec FR Chin Seal for PX5™ PAPR > GVS Z-link Respirator Chin Seal