Learn More
Thermo Scientific™ Z'-LYTE™ Kinase Assay Kit - Ser/Thr 15 Peptide
A coumarin and fluorescein-labeled peptide substrate which can be used for kinase screening in an antibody-free format.
Brand: Thermo Scientific™ PV3799
Description
A coumarin and fluorescein-labeled peptide substrate which can be used for kinase screening in an antibody-free format. This peptide is based upon EGFR, containing Thr-693.

Specifications
Z'-LYTE™ Kinase Assay Kit - Ser/Thr 15 Peptide, 800 x 20μL Assays, 800 rxns, 405/465nm Excitation, 535nm Emission, Methylcoumarin Label, FRET Technique, Serine/Threonine Kinases Target Enzyme, Peptide-Based Substrate Properties, End Point Readout, | |
Cell viability, proliferation and function, pharma and biopharma, protein assays and analysis, target and lead identification and validation, FRET | |
800 x 20μL | |
Biochemical phospho-specific activity (no antibody) | |
Z'-LYTE™ Kinase Assay Kit - Ser/Thr 15 Peptide | |
384-well plate | |
Methylcoumarin | |
End Point | |
Signal Transduction | |
EGFR residues 688-702, centered on Thr-693 (LVEPLTPSGEAPNQK) | |
MAPK11 | |
MAPK11 |
Z ́-LYTE™ | |
All necessary reagents for performing primary or secondary screens | |
Serine/Threonine Kinase Assay | |
–80°C, Z'-LYTE™ Kinase Assay Kits contain all necessary reagents for performing primary or secondary screens. Most kit components should be stored at -20°C, and some kit components should be stored at -80°C. See COA for specific storage instructions. | |
800 x 20μL, 800 x 20 μL Assays | |
405/465, 535 nm | |
800 Reactions | |
J. Cell. Phys.(1995), v. 162, p434 | |
Dry Ice | |
Peptide-Based Substrate | |
Serine/Threonine Kinases |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.