Learn More
Zeochem™ AG ZEOprep™ Classic
ZEOprep™ is a proven, highly successful packing material providing fast, effective and reproducible separations. Irregular shaped silica gel is used in Gravity, Flash and Medium Pressure Chromatography. Available in a variety of pore and particle sizes, moisture contents plus functionalities.
Brand:  Zeochem™ AG 302081
 View more versions of this product
Product Code. 30048984
  • 911.00 € / 500g
Explore more special offers
Description
Description
  • Irregular Amorphous Silica Gel
  • High Purity
  • European Manufactured
  • Swiss accuracy and consistency
  • Excellent batch to batch reproducibility
  • ISO Certification
  • Very wide portfolio
  • Wide Selection of functionalization options
  • Available with 4 different moisture contents
  • High Chemical and mechanical stability
  • Narrow Particle Size Distribution
  • 20 μum to 500 μum particle sizes available
  • Available in 60 A and 90 A pores sizes
  • High loadability
  • Unsurpassed Service and Support
  • Selection of pack sizes available
  • Suitable for gravity, flash and process
Specifications
Specifications
40 to 63 μm
Chromatography
Silica Gel
NH2P
60 Å
500 g
Silica Gel
Irregular