Learn More
Zeochem™ AG ZEOprep™ Classic
ZEOprep™ is a proven, highly successful packing material providing fast, effective and reproducible separations. Irregular shaped silica gel is used in Gravity, Flash and Medium Pressure Chromatography. Available in a variety of pore and particle sizes, moisture contents plus functionalities.
76.00 € - 5000.00 €
Specifications
Technique Chromatography
Type Silica Gel
Material Silica Gel
Particle Shape Irregular
View More Specs
Products 39
Product Code Brand Pore Size Particle Size Phase Quantity Price Quantity & Availability  
Product Code Brand Pore Size Particle Size Phase Quantity Price Quantity & Availability  
30048966
View Documents
Zeochem™ AG
301989
60 Å 25 to 40 μm Bare Silica 1 kg
101.00 €
1kg
30048948
View Documents
Zeochem™ AG
301988
60 Å 25 to 40 μm Bare Silica 5 kg
370.00 €
5kg
30049033
View Documents
Zeochem™ AG
300231
60 Å 40 to 63 μm Bare Silica 1 kg
85.00 €
1kg
30048995
View Documents
Zeochem™ AG
302097
60 Å 40 to 63 μm Bare Silica 5 kg
315.00 €
5kg
30048957
View Documents
Zeochem™ AG
300190
60 Å 40 to 63 μm C18 100 g
249.00 €
100g
30048967
View Documents
Zeochem™ AG
302074
60 Å 40 to 63 μm C18 500 g
867.00 €
500g
30049024
View Documents
Zeochem™ AG
300191
60 Å 40 to 63 μm C18 1 kg
1335.00 €
1kg
30049031
View Documents
Zeochem™ AG
302089
60 Å 40 to 63 μm C18 5 kg
4895.00 €
5kg
30048965
View Documents
Zeochem™ AG
300186
60 Å 40 to 63 μm Diol 100 g
249.00 €
100g
30048949
View Documents
Zeochem™ AG
302076
60 Å 40 to 63 μm Diol 500 g
911.00 €
500g
30049026
View Documents
Zeochem™ AG
300187
60 Å 40 to 63 μm Diol 1 kg
1400.00 €
1kg
30049002
View Documents
Zeochem™ AG
302091
60 Å 40 to 63 μm Diol 5 kg
4770.00 €
5kg
30048962
View Documents
Zeochem™ AG
300184
60 Å 40 to 63 μm NH2 100 g
249.00 €
100g
30048955
View Documents
Zeochem™ AG
302080
60 Å 40 to 63 μm NH2 500 g
911.00 €
500g
30048997
View Documents
Zeochem™ AG
300185
60 Å 40 to 63 μm NH2 1 kg
1400.00 €
1kg
30048973
View Documents
Zeochem™ AG
302092
60 Å 40 to 63 μm NH2 5 kg
4755.00 €
5kg
30049021
View Documents
Zeochem™ AG
300166
60 Å 40 to 63 μm SAX 100 g
249.00 €
100g
30048988
View Documents
Zeochem™ AG
302082
60 Å 40 to 63 μm SAX 500 g
938.00 €
500g
30049015
View Documents
Zeochem™ AG
300167
60 Å 40 to 63 μm SAX 1 kg
1445.00 €
1kg
30049007
View Documents
Zeochem™ AG
302094
60 Å 40 to 63 μm SAX 5 kg
4915.00 €
5kg
30048983
View Documents
Zeochem™ AG
300177
60 Å 40 to 63 μm SH (Thiol) 100 g
249.00 €
100g
30049005
View Documents
Zeochem™ AG
302083
60 Å 40 to 63 μm SH (Thiol) 500 g
938.00 €
500g
30049030
View Documents
Zeochem™ AG
300178
60 Å 40 to 63 μm SH (Thiol) 1 kg
1445.00 €
1kg
30049022
View Documents
Zeochem™ AG
302095
60 Å 40 to 63 μm SH (Thiol) 5 kg
4915.00 €
5kg
30048991
View Documents
Zeochem™ AG
300182
60 Å 40 to 63 μm NH2P 100 g
249.00 €
100g
30048984
View Documents
Zeochem™ AG
302081
60 Å 40 to 63 μm NH2P 500 g
911.00 €
500g
30049020
View Documents
Zeochem™ AG
300183
60 Å 40 to 63 μm NH2P 1 kg
1400.00 €
1kg
30048963
View Documents
Zeochem™ AG
302093
60 Å 40 to 63 μm NH2P 5 kg
4770.00 €
5kg
30048958
View Documents
Zeochem™ AG
300227
60 Å 60 to 200 μm Bare Silica 1 kg
76.00 €
1kg
30048987
View Documents
Zeochem™ AG
302098
60 Å 60 to 200 μm Bare Silica 5 kg
284.00 €
5kg
30048975
View Documents
Zeochem™ AG
300221
60 Å 200 to 500 μm Bare Silica 1 kg
76.00 €
1kg
30048970
View Documents
Zeochem™ AG
302205
60 Å 200 to 500 μm Bare Silica 5 kg
370.00 €
5kg
30048952
View Documents
Zeochem™ AG
302106
90 Å 40 to 63 μm Bare Silica 5 kg
354.00 €
5kg
30048971
View Documents
Zeochem™ AG
300175
90 Å 40 to 63 μm C18 100 g
249.00 €
100g
30049006
View Documents
Zeochem™ AG
302075
90 Å 40 to 63 μm C18 500 g
955.00 €
500g
30049017
View Documents
Zeochem™ AG
300176
90 Å 40 to 63 μm C18 1 kg
1470.00 €
1kg
30048950
View Documents
Zeochem™ AG
302090
90 Å 40 to 63 μm C18 5 kg
5000.00 €
5kg
30049009
View Documents
Zeochem™ AG
302107
90 Å 60 to 200 μm Bare Silica 1 kg
85.00 €
1kg
30048976
View Documents
Zeochem™ AG
302108
90 Å 60 to 200 μm Bare Silica 5 kg
327.00 €
5kg
Description
Description
  • Irregular Amorphous Silica Gel
  • High Purity
  • European Manufactured
  • Swiss accuracy and consistency
  • Excellent batch to batch reproducibility
  • ISO Certification
  • Very wide portfolio
  • Wide Selection of functionalization options
  • Available with 4 different moisture contents
  • High Chemical and mechanical stability
  • Narrow Particle Size Distribution
  • 20 μum to 500 μum particle sizes available
  • Available in 60 A and 90 A pores sizes
  • High loadability
  • Unsurpassed Service and Support
  • Selection of pack sizes available
  • Suitable for gravity, flash and process
Specifications
Specifications
Chromatography
Silica Gel
Silica Gel
Irregular