Learn More
Zinc acrylate, Thermo Scientific Chemicals
36.28 € - 346.00 €
Chemical Identifiers
CAS | 14643-87-9 |
---|---|
Molecular Formula | C6H6O4Zn |
Molecular Weight (g/mol) | 207.49 |
MDL Number | MFCD00067208 |
InChI Key | XKMZOFXGLBYJLS-UHFFFAOYSA-L |
Synonym | zinc acrylate, zinc diacrylate, 2-propenoic acid, zinc salt, acrylic acid, zinc salt, 2-propenoic acid, zinc salt 2:1, zinc prop-2-enoate, 79-10-7 parent, acmc-20aj8t, dsstox_cid_7768, dsstox_rid_78559 |
PubChem CID | 159747 |
IUPAC Name | zinc;prop-2-enoate |
SMILES | [Zn++].[O-]C(=O)C=C.[O-]C(=O)C=C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11358928
|
Thermo Scientific Alfa Aesar
041780.18 |
50 g |
36.28 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11378928
|
Thermo Scientific Alfa Aesar
041780.30 |
250 g |
116.00 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11368928
|
Thermo Scientific Alfa Aesar
041780.A1 |
1 kg |
346.00 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The primary use of Zinc Acrylate is in the production of golf balls. Other uses are primarily in the production of specialty rubber products such as belts and hoses for automobiles. Also used as a tissue adhesive used for adherence of tissue to tissue or tissue to non-tissue for prostheses in vitro.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
14643-87-9 | |
207.49 | |
XKMZOFXGLBYJLS-UHFFFAOYSA-L | |
159747 | |
[Zn++].[O-]C(=O)C=C.[O-]C(=O)C=C |
C6H6O4Zn | |
MFCD00067208 | |
zinc acrylate, zinc diacrylate, 2-propenoic acid, zinc salt, acrylic acid, zinc salt, 2-propenoic acid, zinc salt 2:1, zinc prop-2-enoate, 79-10-7 parent, acmc-20aj8t, dsstox_cid_7768, dsstox_rid_78559 | |
zinc;prop-2-enoate |
Specifications
14643-87-9 | |
1.6 g/mL | |
MFCD00067208 | |
zinc acrylate, zinc diacrylate, 2-propenoic acid, zinc salt, acrylic acid, zinc salt, 2-propenoic acid, zinc salt 2:1, zinc prop-2-enoate, 79-10-7 parent, acmc-20aj8t, dsstox_cid_7768, dsstox_rid_78559 | |
XKMZOFXGLBYJLS-UHFFFAOYSA-L | |
zinc;prop-2-enoate | |
159747 | |
Powder |
∼250°C | |
C6H6O4Zn | |
UN3077 | |
Soluble completely in acrylic acid and water (as a salt). Insoluble as a polymer. | |
[Zn++].[O-]C(=O)C=C.[O-]C(=O)C=C | |
207.49 | |
207.5 | |
Zinc acrylate |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P260-P264b-P270-P271-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c
H302-H314-H317-H335
missing translation for 'dotInformation' : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
missing translation for 'einecsNumber' : 238-692-3
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.