missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc Chloride, Reagent grade, ≥98%, Honeywell™
Reagent Grade,≥98%
19.37 € - 133.07 €
Chemical Identifiers
CAS | 7646-85-7 |
---|---|
Molecular Formula | Cl2Zn |
Molecular Weight (g/mol) | 136.28 |
MDL Number | MFCD00011295 |
InChI Key | JIAARYAFYJHUJI-UHFFFAOYSA-L |
Synonym | zinc chloride, zinc dichloride, zinc chloride zncl2, zinc butter, zinc chloride fume, zinc ii chloride, zinkchloride, zintrace, zinc chloride, anhydrous, zine dichloride |
PubChem CID | 5727 |
ChEBI | CHEBI:49976 |
IUPAC Name | dichlorozinc |
SMILES | Cl[Zn]Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15651820
|
Honeywell Research Chemicals
208086-5G |
5 g | Poly Bottle | N/A | |||||
15621820
|
Honeywell Research Chemicals
208086-100G |
100 g | Poly Bottle | N/A | |||||
15641820
|
Honeywell Research Chemicals
208086-500G |
500 g | Poly Bottle | N/A | |||||
15631820
|
Honeywell Research Chemicals
208086-1KG |
1 kg | Poly Bottle | N/A | |||||
Description
- ≥95% for general laboratory use.
Chemical Identifiers
7646-85-7 | |
136.28 | |
JIAARYAFYJHUJI-UHFFFAOYSA-L | |
5727 | |
dichlorozinc |
Cl2Zn | |
MFCD00011295 | |
zinc chloride, zinc dichloride, zinc chloride zncl2, zinc butter, zinc chloride fume, zinc ii chloride, zinkchloride, zintrace, zinc chloride, anhydrous, zine dichloride | |
CHEBI:49976 | |
Cl[Zn]Cl |
Specifications
293°C | |
White | |
Cl2Zn | |
MFCD00011295 | |
zinc chloride, zinc dichloride, zinc chloride zncl2, zinc butter, zinc chloride fume, zinc ii chloride, zinkchloride, zintrace, zinc chloride, anhydrous, zine dichloride | |
Cl[Zn]Cl | |
136.28 | |
CHEBI:49976 | |
≥98% | |
1mmHg (428°C) | |
Zinc Chloride |
7646-85-7 | |
Solid | |
ZnCl2 | |
UN2331 | |
JIAARYAFYJHUJI-UHFFFAOYSA-L | |
dichlorozinc | |
5727 | |
136.3g/mol | |
Reagent | |
2.93g/cu.cm |
Safety and Handling
P260-P280-P301 + P312 + P330-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338
H302-H314-H410
missing translation for 'dotInformation' : 8/PGIII
missing translation for 'einecsNumber' : 231-592-0
missing translation for 'rtecsNumber' : ZH1400000
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title