missing translation for 'onlineSavingsMsg'
Learn More
Zinc, Oil based standard solution, Specpure™ Zn 5000μg/g, Thermo Scientific Chemicals Product Code.: 15486475

Zinc, Oil based standard solution, Specpure™ Zn 5000μg/g, Thermo Scientific Chemicals

Product Code. 15486475
50 g
Click to view available options
Quantity:
50 g
This item is not returnable. View return policy

Product Code. 15486475

Brand: Thermo Scientific Alfa Aesar 042453.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 50 g
MDL Number MFCD00011291
Physical Form Liquid
Grade Specpure™
Chemical Name or Material Zinc, Oil based standard solution, Zn 5000μg/g

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number1502179 Certificate TypeCertificate of Analysis Date19/11/2024 Catalog Number042453.18
Lot Number1490165 Certificate TypeCertificate of Analysis Date15/10/2024 Catalog Number042453.18
Lot Number1471955 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog Number042453.18
Lot Number289802 Certificate TypeCertificate of Analysis Date21/05/2022 Catalog Number042453.18
Lot Number169499AD Certificate TypeCertificate of Analysis Date22/11/2021 Catalog Number042453.18

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Zinc, Oil based standard solution, Specpure™ Zn 5000μg/g, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.