Learn More
Zirconium carbide, 99.5% (metals basis excluding Hf), Hf< 200ppm, Thermo Scientific Chemicals
93.47 € - 341.70 €
Chemical Identifiers
CAS | 12070-14-3 |
---|---|
Molecular Formula | CZr |
Molecular Weight (g/mol) | 103.24 |
MDL Number | MFCD00049640 |
InChI Key | MHMXYYDKIQTOHA-UHFFFAOYSA-N |
IUPAC Name | methanidylidynezirconiumylium |
SMILES | [C-]#[Zr+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11388707
|
Thermo Scientific Alfa Aesar
035808.18 |
50 g |
93.47 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11378707
|
Thermo Scientific Alfa Aesar
035808.30 |
250 g |
341.70 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Zirconium carbide commercially used in tool bits for cutting tools. It is used as an abrasive, in cladding, in cermets and incandescent filaments. It finds application in the coating of uranium dioxide and thorium dioxide particles of nuclear fuel as well as in thermo-photovoltaic radiators and field emitter tips and arrays. It is involved in the usage as refractory coatings in nuclear reactors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
12070-14-3 | |
103.24 | |
MHMXYYDKIQTOHA-UHFFFAOYSA-N | |
[C-]#[Zr+] |
CZr | |
MFCD00049640 | |
methanidylidynezirconiumylium |
Specifications
3540°C | |
Gray | |
CZr | |
UN3178 | |
MHMXYYDKIQTOHA-UHFFFAOYSA-N | |
methanidylidynezirconiumylium | |
103.23 | |
Odorless | |
(metals basis excluding Hf), Hf< 200ppm |
12070-14-3 | |
Powder | |
MFCD00049640 | |
Soluble in concentrated sulfuric acid,hydrofluoric acid and nitric acid. Insoluble in water. | |
[C-]#[Zr+] | |
103.24 | |
99.5% | |
6.73 g/mL | |
Zirconium carbide |
Safety and Handling
GHS H Statement
H228
Flammable solid.
P210-P240-P241-P280-P370+P378q
H228
missing translation for 'dotInformation' : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLID, INORGANIC, N.O.S.
missing translation for 'einecsNumber' : 235-125-1
missing translation for 'rtecsNumber' : ZH7155000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only