missing translation for 'onlineSavingsMsg'
Learn More

Zirconium silicide, 99.5% (metals basis excluding Hf), Hf 4% max, Thermo Scientific Chemicals

122.61 € - 341.70 €

Chemical Identifiers

CAS 12039-90-6
Molecular Formula H4Si2Zr
Molecular Weight (g/mol) 151.43
MDL Number MFCD00151547
InChI Key ZSONHIWWHDAZQO-UHFFFAOYSA-N
Synonym zirconium silicide, zirconium silicide zrsi2, zirconium silicide, zrsi2, $l^ 5-silanylidenezirconiumylidene-$l^ 5-silane
PubChem CID 6336892
IUPAC Name bis($l^{2}-silanylidene)zirconium
SMILES [SiH2]=[Zr]=[SiH2]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11373408
View Documents
Thermo Scientific Alfa Aesar
036322.14
25 g
122.61 €
25g
Please to purchase this item. Need a web account? Register with us today!
11363408
View Documents
Thermo Scientific Alfa Aesar
036322.22
100 g
341.70 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Zirconium silicate is used for manufacturing refractory materials. It is also used in production of some ceramics, enamels, and ceramic glazes. In enamels and glazes it serves as an opacifier. zirconium silicate is as beads for milling and grinding.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

12039-90-6
151.43
ZSONHIWWHDAZQO-UHFFFAOYSA-N
6336892
[SiH2]=[Zr]=[SiH2]
H4Si2Zr
MFCD00151547
zirconium silicide, zirconium silicide zrsi2, zirconium silicide, zrsi2, $l^ 5-silanylidenezirconiumylidene-$l^ 5-silane
bis($l^{2}-silanylidene)zirconium

Specifications

12039-90-6
Powder
MFCD00151547
Insoluble in water. It is insoluble in acids,alkali and aqua regia.
[SiH2]=[Zr]=[SiH2]
151.43
147.39
Odorless
(metals basis excluding Hf), Hf 4%
Gray
H4Si2Zr
zirconium silicide, zirconium silicide zrsi2, zirconium silicide, zrsi2, $l^ 5-silanylidenezirconiumylidene-$l^ 5-silane
ZSONHIWWHDAZQO-UHFFFAOYSA-N
bis($l^{2}-silanylidene)zirconium
6336892
99.5%
4.88 g/mL
Zirconium silicide
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 234-911-1

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only