accessibility menu, dialog, popup

Test

CAS RN 10196-18-6

CAS RN 10196-18-6

IUPAC Name:
Synonyms:
Molecular Weight (g/mol): 287.66
Molecular Formula: C6H26N2O6Zn
InChi Key: PZDYRUXUUCEJGL-UHFFFAOYSA-N
SMILES: C.C.C.C.C.C.[Zn].O[N+]([O-])=O.O[N+]([O-])=O
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (7)
  • (1)
Physical Form 
  • (3)
  • (3)
  • (2)
  • (2)
  • (7)
Percent Purity 
  • (2)
  • (9)
  • (2)
Grade 
  • (2)
  • (3)
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (5)
  • (2)
  • (3)
  • (8)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (7)
  • (1)
  • (3)
  • (3)
  • (2)
  • (2)
  • (7)
  • (2)
  • (9)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)