Learn More
1,3-Dibromoacetone, 95%, Thermo Scientific Chemicals
34.87 € - 313.56 €
Chemical Identifiers
CAS | 816-39-7 |
---|---|
Molecular Formula | C3H4Br2O |
Molecular Weight (g/mol) | 215.872 |
MDL Number | MFCD00013540 |
InChI Key | LQQKDSXCDXHLLF-UHFFFAOYSA-N |
Synonym | 1,3-dibromoacetone, 1,3-dibromo-2-propanone, 2-propanone, 1,3-dibromo, 1,3-dibromopropanone, bromomethyl ketone, bromomethylketone, sgqdllrxbrwsp@, 1,3-dibromacetone, acmc-20ap4e, 2-propanone,3-dibromo |
PubChem CID | 69952 |
IUPAC Name | 1,3-dibromopropan-2-one |
SMILES | C(C(=O)CBr)Br |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15497747
|
Thermo Scientific Alfa Aesar
H37325.03 |
1 g |
34.87 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15407757
|
Thermo Scientific Alfa Aesar
H37325.06 |
5 g |
72.56 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15417757
|
Thermo Scientific Alfa Aesar
H37325.14 |
25 g |
313.56 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
1,3-Dibromoacetone is used as an antibiotic and antimicrobial agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1,3-Dibromoacetone is used as an antibiotic and antimicrobial agent.
Solubility
Soluble in acetone, chloroform, dichloromethane and methanol.
Notes
Air and moisture sensitive. Incompatible with oxidizing agents and heat.
Chemical Identifiers
816-39-7 | |
215.872 | |
LQQKDSXCDXHLLF-UHFFFAOYSA-N | |
69952 | |
C(C(=O)CBr)Br |
C3H4Br2O | |
MFCD00013540 | |
1,3-dibromoacetone, 1,3-dibromo-2-propanone, 2-propanone, 1,3-dibromo, 1,3-dibromopropanone, bromomethyl ketone, bromomethylketone, sgqdllrxbrwsp@, 1,3-dibromacetone, acmc-20ap4e, 2-propanone,3-dibromo | |
1,3-dibromopropan-2-one |
Specifications
816-39-7 | |
White to Yellow | |
102°C (216°F) | |
MFCD00013540 | |
1740389 | |
1,3-dibromoacetone, 1,3-dibromo-2-propanone, 2-propanone, 1,3-dibromo, 1,3-dibromopropanone, bromomethyl ketone, bromomethylketone, sgqdllrxbrwsp@, 1,3-dibromacetone, acmc-20ap4e, 2-propanone,3-dibromo | |
LQQKDSXCDXHLLF-UHFFFAOYSA-N | |
1,3-dibromopropan-2-one | |
69952 | |
98% | |
1,3-Dibromoacetone |
29°C to 30°C | |
97°C to 98°C (21 mmHg) | |
C3H4Br2O | |
UN1759 | |
Moisture sensitive | |
Soluble in acetone,chloroform,dichloromethane and methanol. | |
C(C(=O)CBr)Br | |
215.872 | |
215.87 | |
Crystals or Fused Solid |
Safety and Handling
GHS H Statement
H314-H318-H317
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause an allergic skin reaction.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packaging Group: II; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.
missing translation for 'einecsNumber' : 212-430-8
missing translation for 'tsca' : Yes
missing translation for 'storageNote' : 2° to 8°C
Recommended Storage : Keep cold; Store under Argon
- 1,3-Dibromoacetone
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : 1,3-Dibromoacetone
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.