missing translation for 'onlineSavingsMsg'
Learn More
1,3-Dithiane, 97%, Thermo Scientific Chemicals Product Code.: 15760819

1,3-Dithiane, 97%, Thermo Scientific Chemicals

Product Code. 15760819
100 g, Glass bottle
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 15760819

Brand: Thermo Scientific Acros 169511000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 505-23-7
Molecular Formula C4H8S2
Molecular Weight (g/mol) 120.23
MDL Number MFCD00006654
InChI Key WQADWIOXOXRPLN-UHFFFAOYSA-N
Synonym 1,3-dithiacyclohexane, m-dithiane, dithiane-1,3, 1,3-dithian, dithiane-1,3 french, 1,3 dithiane, ccris 6777, m-dithiane, 8ci, 1,3 dithiocyclohexane, acmc-209knkShow MoreShow Less
PubChem CID 10451
ChEBI CHEBI:47845
IUPAC Name 1,3-dithiane
SMILES C1CSCSC1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 49°C to 54°C
Color Beige to White
Density 90g/mL
Flash Point 90°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 100 g
Fieser 02,182; 03,135; 04,216; 05,287; 06,248; 07,142; 14,157; 16,161; 01,208; 02,108; 12,158; 15,112
Formula Weight 120.23
Percent Purity 97%
Physical Form Crystals, Needles or Powder
Chemical Name or Material 1, 3-Dithiane
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0436354 Certificate TypeCertificate of Analysis Date30/01/2025 Catalog Number169510250,169511000,169510050
Lot NumberA0436354 Certificate TypeCertificate of Analysis Date16/11/2023 Catalog Number169510250,169511000,169510050
Lot NumberA0455473 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number169510250,169511000,169510050
Lot NumberA0404107 Certificate TypeCertificate of Analysis Date12/12/2022 Catalog Number169510250,169511000,169510050
Lot NumberA0406341 Certificate TypeCertificate of Analysis Date08/03/2021 Catalog Number169510250,169511000,169510050

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1,3-Dithiane, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.