missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals 1,6-Anhydro-β-D-glucopyranose, 99+% Product Code.: 10279153

Thermo Scientific Chemicals 1,6-Anhydro-β-D-glucopyranose, 99+%

Product Code. 10279153
5 g, Glass bottle
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10279153

Brand: Thermo Scientific Chemicals 270950050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 498-07-7
Molecular Formula C6H10O5
Molecular Weight (g/mol) 162.14
MDL Number MFCD00063248
InChI Key TWNIBLMWSKIRAT-UHFFFAOYNA-N
Synonym 1,6-anhydro-beta-d-glucopyranose, levoglucosan, leucoglucosan, 1,6-anhydro-beta-d-glucose, 1,6-anhydro-beta-glucopyranose, glucosan, 1,6-anhydroglucose, unii-5132n17fsd, anhydroglucose, 1,6-anhydro-d-glucoseShow MoreShow Less
PubChem CID 2724705
ChEBI CHEBI:30997
SMILES OC1C2COC(O2)C(O)C1O

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 170.0°C to 180.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 99% min. (ELSD) (HPLC)
Packaging Glass bottle
Quantity 5 g
Specific Rotation Condition − 66.00 (20.00°C c=2,water)
Specific Rotation − 66.00
Solubility Information (5% in water) Clear colorless
IUPAC Name 6,8-dioxabicyclo[3.2.1]octane-2,3,4-triol
Formula Weight 162.14
Percent Purity 99+%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 1,6-Anhydro-β-D-glucopyranose
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0448673 Certificate TypeCertificate of Analysis Date05/01/2023 Catalog Number270950010,270950050
Lot NumberA0320539 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number270950010,270950050
Lot NumberA0409815 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number270950010,270950050
Lot NumberA0417559 Certificate TypeCertificate of Analysis Date12/09/2022 Catalog Number270950010,270950050

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals 1,6-Anhydro-β-D-glucopyranose, 99+% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.