missing translation for 'onlineSavingsMsg'
Learn More
1-Bromo-2-iodotetrafluoroethane, 97%, stab. with copper, Thermo Scientific Chemicals Product Code.: 11460397

1-Bromo-2-iodotetrafluoroethane, 97%, stab. with copper, Thermo Scientific Chemicals

Product Code. 11460397
5 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11460397

Brand: Thermo Scientific Alfa Aesar B21771.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 421-70-5
Molecular Formula C2BrF4I
Molecular Weight (g/mol) 306.82
MDL Number MFCD00042136
InChI Key ZYNPYKGTNSXKPI-UHFFFAOYSA-N
Synonym 1-bromo-2-iodotetrafluoroethane, 1-bromotetrafluoro-2-iodoethane, unii-e75xu1n0mr, e75xu1n0mr, ethane, 1-bromo-1,1,2,2-tetrafluoro-2-iodo, ccris 9437, acmc-1amr3, 1-bromo-2-iodoperfluoroethane, 1-iodo-2-bromotetrafluoroethane, 2-iodo-1-bromotetrafluoroethaneShow MoreShow Less
PubChem CID 67902
IUPAC Name 1-bromo-1,1,2,2-tetrafluoro-2-iodoethane
SMILES FC(F)(Br)C(F)(F)I

Specifications

Density 1.648
Boiling Point 78°C
Refractive Index 1.433
Quantity 5 g
Beilstein 1688466
Sensitivity Light sensitive
Formula Weight 306.82
Percent Purity 97%
Chemical Name or Material 1-Bromo-2-iodotetrafluoroethane, stabilized with copper

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10172543 Certificate TypeCertificate of Analysis Date28/06/2023 Catalog NumberB21771,B21771.22,B21771.14,B21771.06
Lot Number10172543 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB21771.22,B21771.14,B21771.06
Lot Number10158950 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB21771.22,B21771.14,B21771.06
Lot Number10161276 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB21771.22,B21771.14,B21771.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Bromo-2-iodotetrafluoroethane, 97%, stab. with copper, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.