missing translation for 'onlineSavingsMsg'
Learn More
1-Bromobutane, 98+%, Thermo Scientific Chemicals Product Code.: 11413464

1-Bromobutane, 98+%, Thermo Scientific Chemicals

Product Code. 11413464
10,000 g
Click to view available options
Quantity:
10,000 g
2500 g
500 g
This item is not returnable. View return policy

Product Code. 11413464

Brand: Thermo Scientific Alfa Aesar A10696.0C

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

1-Bromobutane is used as an intermediate in organic synthesis and as a solvent for cleaning and degreasing. It acts as an alkylating agent as well as to prepare organometallic compounds such as n-butyllithium. It is also involved in the synthesis of procaine and tetracaine. It reacts with magnesium metal to prepare the Grignard reagent, which is used to form carbon-carbon bonds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromobutane is used as an intermediate in organic synthesis and as a solvent for cleaning and degreasing. It acts as an alkylating agent as well as to prepare organometallic compounds such as n-butyllithium. It is also involved in the synthesis of procaine and tetracaine. It reacts with magnesium metal to prepare the Grignard reagent, which is used to form carbon-carbon bonds.

Solubility
Miscible with ethanol and diethyl ether. Immiscible with water.

Notes
Incompatible with strong oxidizing agents, strong bases, magnesium, potassium, sodium and sodium oxides.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-65-9
Molecular Formula C4H9Br
Molecular Weight (g/mol) 137.02
MDL Number MFCD00000260
InChI Key MPPPKRYCTPRNTB-UHFFFAOYSA-N
Synonym butyl bromide, n-butyl bromide, bromobutane, butane, 1-bromo, 1-butyl bromide, butane, bromo, n-butylbromide, 1-bromo-butane, unii-sav6y78u3d, ccris 831Show MoreShow Less
PubChem CID 8002
IUPAC Name 1-bromobutane
SMILES CCCCBr

Specifications

Melting Point -112°C
Density 1.275
Boiling Point 101°C to 102°C
Flash Point 20°C (68°F)
Assay Percent Range >98%
Odor Characteristic
Linear Formula CH3(CH2)3Br
Refractive Index 1.44
Quantity 10,000 g
UN Number UN1126
Beilstein 1098260
Merck Index 14,1553
Solubility Information Miscible with ethanol and diethyl ether. Immiscible with water.
Formula Weight 137.03
Percent Purity ≥98%
Chemical Name or Material 1-Bromobutane
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10252083 Certificate TypeCertificate of Analysis Date15/08/2024 Catalog NumberA10696.0E,A10696.0C,A10696.36
Lot Number10252082 Certificate TypeCertificate of Analysis Date15/08/2024 Catalog NumberA10696.0E,A10696.0C,A10696.36
Lot Number10226499 Certificate TypeCertificate of Analysis Date05/12/2023 Catalog NumberA10696.0E,A10696.0C,A10696.36
Lot Number10245504 Certificate TypeCertificate of Analysis Date26/01/2023 Catalog NumberA10696.0E,A10696.0C,A10696.36
Lot Number10242084 Certificate TypeCertificate of Analysis Date18/07/2022 Catalog NumberA10696.0E,A10696.0C,A10696.36
compliance-icons compliance-icons
Product Identifier
  • 1-Bromobutane
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
Indicazioni di pericolo
  • H225-Highly flammable liquid and vapour.
  • H315-Causes skin irritation.
Consigli di prudenza
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
Informazioni supplementari
  • MIXTURE LIST-Contains : 1-Bromobutane

RUO – Research Use Only

Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto
1-Bromobutane, 98+%, Thermo Scientific Chemicals >

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato