missing translation for 'onlineSavingsMsg'
Learn More

1-Bromobutane, 98+%, Thermo Scientific Chemicals

43.92 € - 378.89 €

Chemical Identifiers

CAS 109-65-9
Molecular Formula C4H9Br
Molecular Weight (g/mol) 137.02
MDL Number MFCD00000260
InChI Key MPPPKRYCTPRNTB-UHFFFAOYSA-N
Synonym butyl bromide, n-butyl bromide, bromobutane, butane, 1-bromo, 1-butyl bromide, butane, bromo, n-butylbromide, 1-bromo-butane, unii-sav6y78u3d, ccris 831
PubChem CID 8002
IUPAC Name 1-bromobutane
SMILES CCCCBr
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11493454
View Documents
Thermo Scientific Alfa Aesar
A10696.36
500 g
43.92 €
500g
Please to purchase this item. Need a web account? Register with us today!
11403464
View Documents
Thermo Scientific Alfa Aesar
A10696.0E
2500 g
121.61 €
2500g
Please to purchase this item. Need a web account? Register with us today!
11413464
View Documents
Thermo Scientific Alfa Aesar
A10696.0C
10,000 g
378.89 €
10000g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

1-Bromobutane is used as an intermediate in organic synthesis and as a solvent for cleaning and degreasing. It acts as an alkylating agent as well as to prepare organometallic compounds such as n-butyllithium. It is also involved in the synthesis of procaine and tetracaine. It reacts with magnesium metal to prepare the Grignard reagent, which is used to form carbon-carbon bonds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromobutane is used as an intermediate in organic synthesis and as a solvent for cleaning and degreasing. It acts as an alkylating agent as well as to prepare organometallic compounds such as n-butyllithium. It is also involved in the synthesis of procaine and tetracaine. It reacts with magnesium metal to prepare the Grignard reagent, which is used to form carbon-carbon bonds.

Solubility
Miscible with ethanol and diethyl ether. Immiscible with water.

Notes
Incompatible with strong oxidizing agents, strong bases, magnesium, potassium, sodium and sodium oxides.
Specifications

Chemical Identifiers

109-65-9
137.02
MPPPKRYCTPRNTB-UHFFFAOYSA-N
8002
CCCCBr
C4H9Br
MFCD00000260
butyl bromide, n-butyl bromide, bromobutane, butane, 1-bromo, 1-butyl bromide, butane, bromo, n-butylbromide, 1-bromo-butane, unii-sav6y78u3d, ccris 831
1-bromobutane

Specifications

109-65-9
1.275 g/mL
20°C (68°F)
C4H9Br
1.44
UN1126
14,1553
Miscible with ethanol and diethyl ether. Immiscible with water.
CCCCBr
137.02
137.03
1-Bromobutane
°C to 112°C
101°C to 102°C
>98%
CH3(CH2)3Br
MFCD00000260
1098260
butyl bromide, n-butyl bromide, bromobutane, butane, 1-bromo, 1-butyl bromide, butane, bromo, n-butylbromide, 1-bromo-butane, unii-sav6y78u3d, ccris 831
MPPPKRYCTPRNTB-UHFFFAOYSA-N
1-bromobutane
8002
≥98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H315-H319-H335

P210-P233-P240-P241-P242-P243-P264b-P280-P303+P361+P353-P305+P351+P338-P332+P313-P363-P370+P378q-P501c

H225-H315-H319

missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: 1-BROMOBUTANE

missing translation for 'einecsNumber' : 203-691-9

missing translation for 'rtecsNumber' : EJ6225000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • 1-Bromobutane

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H361fd-Suspected of damaging fertility. Suspected of damaging the unborn child.
  • H372-Causes damage to organs.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.

Supplemental information
  • MIXTURE LIST-Contains : Butyl bromide

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.