missing translation for 'onlineSavingsMsg'
Learn More
1-Bromopentane, 99%, Thermo Scientific Chemicals Product Code.: 11499762

1-Bromopentane, 99%, Thermo Scientific Chemicals

Product Code. 11499762
250 g
Click to view available options
Quantity:
250 g
1000 g
5000 g
This item is not returnable. View return policy

Product Code. 11499762

Brand: Thermo Scientific Alfa Aesar A10821.30

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

1-Bromopentane is used in a multitude of organic synthesis. It is used in iron-catalyzed alkylations of aromatic Grignard reagents via cross coupling.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromopentane is used in a multitude of organic synthesis. It is used in iron-catalyzed alkylations of aromatic Grignard reagents via cross coupling.

Solubility
Insoluble in water and soluble in alcohol. Miscible in diethyl ether.

Notes
Store away from oxidizing agents and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-53-2
Molecular Formula C5H11Br
Molecular Weight (g/mol) 151.047
MDL Number MFCD00000267
InChI Key YZWKKMVJZFACSU-UHFFFAOYSA-N
Synonym n-amyl bromide, pentyl bromide, amyl bromide, pentane, 1-bromo, n-pentyl bromide, 1-pentyl bromide, bromopentane, 1-bromo-pentane, 1-pentylbromide, unii-z2s4r599p0Show MoreShow Less
PubChem CID 8057
IUPAC Name 1-bromopentane
SMILES CCCCCBr

Specifications

Melting Point -95°C
Density 1.215
Boiling Point 127°C to 129°C
Flash Point 31°C (87°F)
Odor Ether-like
Refractive Index 1.4436
Quantity 250 g
UN Number UN1993
Beilstein 1730981
Merck Index 14,602
Solubility Information Insoluble in water and soluble in alcohol. Miscible in diethyl ether.
Formula Weight 151.05
Percent Purity 99%
Chemical Name or Material 1-Bromopentane
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10244392 Certificate TypeCertificate of Analysis Date31/10/2022 Catalog NumberA10821.0B,A10821.0I,A10821.30
Lot Number10239273 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA10821.0B,A10821.0I,A10821.30
Lot Number5002N03N Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA10821.0B,A10821.0I,A10821.30
Lot Number10221246 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA10821.0B,A10821.0I,A10821.30
Lot NumberG5479B Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA10821.0B,A10821.0I,A10821.30
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 1-Bromopentane
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : 1-Bromopentane

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Bromopentane, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.