Learn More
2-Aminopyrimidine-5-boronic acid pinacol ester, 96%, Thermo Scientific™
73.20 € - 885.00 €
Chemical Identifiers
CAS | 402960-38-7 |
---|---|
Molecular Formula | C10H16BN3O2 |
Molecular Weight (g/mol) | 221.067 |
MDL Number | MFCD06795685 |
InChI Key | BPQVMIDUTRJYSC-UHFFFAOYSA-N |
Synonym | 2-aminopyrimidine-5-boronic acid pinacol ester, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidin-2-amine, 2-aminopyrimidine-5-boronic acid, pinacol ester, 5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidin-2-amine, 2-amino-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidine, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-pyrimidin-2-amine, 2-aminopyrimidin-5-yl boronic acid pinacol ester, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2-pyrimidinamine, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2-pyrimidinylamine, 2-2-aminopyrimidin-5-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane |
PubChem CID | 17750212 |
IUPAC Name | 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidin-2-amine |
SMILES | B1(OC(C(O1)(C)C)(C)C)C2=CN=C(N=C2)N |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications2-Aminopyrimidine-5-boronic acid pinacol ester is used in the preparation of phosphoinositide-3-kinases (PI3K) inhibitors.
Solubility
Slightly soluble in water. Soluble in chloroform.
Notes
Incompatible with strong oxidizing agents, reducing agents and acids. Store at 4-2°C.
Chemical Identifiers
402960-38-7 | |
221.067 | |
BPQVMIDUTRJYSC-UHFFFAOYSA-N | |
17750212 | |
B1(OC(C(O1)(C)C)(C)C)C2=CN=C(N=C2)N |
C10H16BN3O2 | |
MFCD06795685 | |
2-aminopyrimidine-5-boronic acid pinacol ester, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidin-2-amine, 2-aminopyrimidine-5-boronic acid, pinacol ester, 5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidin-2-amine, 2-amino-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidine, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-pyrimidin-2-amine, 2-aminopyrimidin-5-yl boronic acid pinacol ester, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2-pyrimidinamine, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2-pyrimidinylamine, 2-2-aminopyrimidin-5-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane | |
5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidin-2-amine |
Specifications
402960-38-7 | |
C10H16BN3O2 | |
2-aminopyrimidine-5-boronic acid pinacol ester, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidin-2-amine, 2-aminopyrimidine-5-boronic acid, pinacol ester, 5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidin-2-amine, 2-amino-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidine, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-pyrimidin-2-amine, 2-aminopyrimidin-5-yl boronic acid pinacol ester, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2-pyrimidinamine, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2-pyrimidinylamine, 2-2-aminopyrimidin-5-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane | |
BPQVMIDUTRJYSC-UHFFFAOYSA-N | |
5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidin-2-amine | |
17750212 | |
96% |
206°C to 208°C | |
MFCD06795685 | |
Slightly soluble in water. Soluble in chloroform. | |
B1(OC(C(O1)(C)C)(C)C)C2=CN=C(N=C2)N | |
221.067 | |
221.06 | |
2-Aminopyrimidine-5-boronic acid pinacol ester |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Keep cold; Store under Argon
Your input is important to us. Please complete this form to provide feedback related to the content on this product.