missing translation for 'onlineSavingsMsg'
Learn More
2-Butanone, +99%, Extra Pure, Thermo Scientific Chemicals Product Code.: 10306510

2-Butanone, +99%, Extra Pure, Thermo Scientific Chemicals

Product Code. 10306510
1 L, Glass Bottle
Click to view available options
Quantity:
25 mL
1 L
2.5 L
25 L
Packaging:
Glass Bottle
Metal Drum
This item is not returnable. View return policy

Product Code. 10306510

Brand: Thermo Scientific Acros 149670010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-93-3
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00011648
InChI Key ZWEHNKRNPOVVGH-UHFFFAOYSA-N
Synonym 2-butanone, methyl ethyl ketone, butanone, ethyl methyl ketone, methylethyl ketone, meetco, methyl acetone, methylethylketone, 3-butanone, ethylmethylketonShow MoreShow Less
PubChem CID 6569
ChEBI CHEBI:28398
IUPAC Name butan-2-one
SMILES CCC(C)=O

Specifications

Melting Point -87.0°C
CAS Min % 99.0
Color Colorless
CAS Max % 100.0
Density 0.8060g/mL
Boiling Point 80.0°C
Quantity 1 L
Flash Point −7°C
Infrared Spectrum Authentic
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.3780 to 1.3800
Assay Percent Range 99+%
Residue after Evaporation 0.005% max.
Specific Gravity 0.806
Linear Formula C2H5COCH3
Vapor Pressure Vapor Pressure: 105mbar at 20°C
Viscosity 0.42 mPa.s (15°C)
Beilstein 01, IV, 3243
Water 0.2% max.
Fieser 01,678
Merck Index 15, 6143
Solubility Information Solubility in water: 290g/L (20°C). Other solubilities: soluble in benzene, alcohol and ether, miscible with oils
Formula Weight 72.11
Percent Purity ≥99%
Grade Extra Pure
Chemical Name or Material 2-Butanone
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number1987599 Certificate TypeCertificate of Analysis Date27/01/2025 Catalog Number149670025,149670250,149670251,149670010
Lot Number1994040 Certificate TypeCertificate of Analysis Date27/01/2025 Catalog Number149670025,149670250,149670251,149670010
Lot Number1997720 Certificate TypeCertificate of Analysis Date27/01/2025 Catalog Number149670025,149670250,149670251,149670010
Lot Number2418538 Certificate TypeCertificate of Analysis Date27/01/2025 Catalog Number149670025,149670250,149670251,149670010
Lot Number2106844 Certificate TypeCertificate of Analysis Date27/01/2025 Catalog Number149670025,149670250,149670251,149670010
compliance-icons compliance-icons
Product Identifier
  • 2-Butanone
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Butanone, +99%, Extra Pure, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.