missing translation for 'onlineSavingsMsg'
Learn More
2-Methyl-1,3-dithiane, 97%, Thermo Scientific™ Product Code.: 11936861

2-Methyl-1,3-dithiane, 97%, Thermo Scientific™

Product Code. 11936861
1 mL, Glass bottle
Click to view available options
Quantity:
1 mL
5 mL
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 11936861

Brand: Thermo Scientific Acros 443260010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 6007-26-7
InChI Key KXROTPXCYDXGSC-UHFFFAOYSA-N
Synonym 1,3-dithiane, 2-methyl, m-dithiane, 2-methyl, 2-methyl-m-dithiane, methyl-2 dithiane-1,3, methyl-2 dithiane-1,3 french, acmc-20anrf, 2-methyl-1,3-dithian, 2-methyl-1,3 dithiane, 5-19-01-00053 beilstein handbook reference, 2-methyl-1,3-dithianeShow MoreShow Less
PubChem CID 22329
IUPAC Name 2-methyl-1,3-dithiane
SMILES CC1SCCCS1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Brown
Density 1.1200g/mL
Boiling Point 66°C (5.0 Torr)
Flash Point 82°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C5H10S2
Quantity 1 mL
Specific Gravity 1.12
Molecular Weight (g/mol) 134.26
Formula Weight 134.26
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-Methyl-1, 3-dithiane
Show More Show Less
Product Identifier
  • 2-Methyl-1,3-dithiane

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Methyl-1,3-dithiane, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.