missing translation for 'onlineSavingsMsg'
Learn More
2-Methylbenzoxazole, 97%, Thermo Scientific Chemicals Product Code.: 11494614

2-Methylbenzoxazole, 97%, Thermo Scientific Chemicals

Product Code. 11494614
25 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11494614

Brand: Thermo Scientific Alfa Aesar A13198.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

2-Methylbenzoxazole has been used in synthesis of bis-styryl dyes. It is also used in medicine and for the synthesis of other organic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methylbenzoxazole has been used in synthesis of bis-styryl dyes. It is also used in medicine and for the synthesis of other organic compounds.

Solubility
Not miscible in water.

Notes
It is sensitive to light. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-21-6
Molecular Formula C8H7NO
Molecular Weight (g/mol) 133.15
MDL Number MFCD00005771
InChI Key DQSHFKPKFISSNM-UHFFFAOYSA-N
Synonym 2-methylbenzoxazole, 2-methylbenzo d oxazole, 2-methylbenzoxazol, benzoxazole, 2-methyl, 2-methyl-benzoxazole, usaf ek-982, 2-methyl-benzooxazole, 2-methyl-4,5-benzoxazole, 2-methylbenzoxazol czech, unii-z0p021v3tiShow MoreShow Less
PubChem CID 7225
ChEBI CHEBI:51603
IUPAC Name 2-methyl-1,3-benzoxazole
SMILES CC1=NC2=CC=CC=C2O1

Specifications

Melting Point 2°C to 4°C
Density 1.12
Boiling Point 200°C to 201°C
Flash Point 85°C (185°F)
Refractive Index 1.55
Quantity 25 g
Beilstein 112297
Sensitivity Light sensitive
Solubility Information Not miscible in water.
Formula Weight 133.15
Percent Purity 98%
Chemical Name or Material 2-Methylbenzoxazole
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberW24E015 Certificate TypeCertificate of Analysis Date18/01/2025 Catalog NumberA13198.22,A13198.14
Lot NumberS20J089 Certificate TypeCertificate of Analysis Date26/06/2023 Catalog NumberA13198.22,A13198.14,A13198
Lot NumberQ10J047 Certificate TypeCertificate of Analysis Date09/05/2023 Catalog NumberA13198.22,A13198.14
Lot Number10243381 Certificate TypeCertificate of Analysis Date17/10/2022 Catalog NumberA13198.22,A13198.14
Lot Number10243160 Certificate TypeCertificate of Analysis Date06/09/2022 Catalog NumberA13198.22,A13198.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Methylbenzoxazole, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.