missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ 2000mL BIOS Swinging Bucket Rotors

The unique 8 x 2000mL (16L) capacity of the Thermo Scientific™ 2000mL BIOS swinging bucket rotors enhance the productivity of your bioprocessing methods.

Brand:  Thermo Scientific™ 75003962

29253.65 EUR valid until 2025-04-30
Use promo code "24119" to get your promotional price.


 View more versions of this product

Product Code. 15550194

  • 37587.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Thermo Scientific™ 2000mL BIOS swinging bucket rotors are designed for use with the Thermo Scientific™ Sorvall™ BIOS 16 bioprocessing centrifuge. The rotor, buckets and bottles which are provided as a set, have been engineered to simplify operation of the centrifuge while maximizing productivity.

Achieve efficient and reliable sample processing with the following features:
  • 6 x 2000mL (12L) capacity at 4,700 max rpm (7,187 x g max RCF) operation;
  • 8 x 2000mL (16L)  capacity at 3,900 max rpm (5,374 x g max RCF) operation
  • Easy-to-use innovative 2000mL bucket and bottle with a wide mouth design requires no bridge and are resistant to 1000 cycles
  • Thermo Scientific™ Eco-Spin™ windshield provides energy savings up to 60% when compared with non-windshielded rotors
  • Compatible with Thermo Scientific™ Auto-Lid™ and Auto-ID™ enabling instant lid opening, storage and rotor identification that saves time and protects the integrity of your samples
  • 2-year warranty

Upgrade to Thermo Scientific™ Dura-Coat™ buckets with nickel-coating to improve protection against corrosion due to moisture, chemicals or alkaline solutions.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Stainless Steel Rotor Body, Aluminum Windshield and Bucket, Polypropylene Bottle
6 x 2000 mL
4,700 rpm
Rotor, Oval Buckets, 2000 mL Bottles
Swinging Bucket Rotor
7,187 x g
Thermo Scientific Sorvall BIOS 16 Centrifuge
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ 2000mL BIOS Swinging Bucket Rotors > 6 x 2000 mL, Oval Buckets. 2000 mL Bottles, 4,700rpm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.