Learn More
Thermo Scientific™ 2000mL BIOS Swinging Bucket Rotors
The unique 8 x 2000mL (16L) capacity of the Thermo Scientific™ 2000mL BIOS swinging bucket rotors enhance the productivity of your bioprocessing methods.
4040.00 € - 46145.00 €
Product Code | Brand | Description | Capacity | Max. Speed | Max. RCF | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Capacity | Max. Speed | Max. RCF | Price | Quantity & Availability | ||
15550194
|
Thermo Scientific™
75003962 |
6 x 2000 mL Swinging Bucket Rotor, Oval Buckets and 2000 mL Bottles | 6 x 2000 mL | 4,700 rpm | 7,187 x g |
36875.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15500204
|
Thermo Scientific™
75003982 |
8 x 2000 mL Swinging Bucket Rotor, Oval Buckets and 2000 mL Bottles | 8 x 2000 mL | 3,900 rpm | 5,374 x g |
46145.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15570194
|
Thermo Scientific™
75003964 |
Oval Bucket (set of 2) |
4040.00 €
Pack of 2 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Thermo Scientific™ 2000mL BIOS swinging bucket rotors are designed for use with the Thermo Scientific™ Sorvall™ BIOS 16 bioprocessing centrifuge. The rotor, buckets and bottles which are provided as a set, have been engineered to simplify operation of the centrifuge while maximizing productivity.
Achieve efficient and reliable sample processing with the following features:- 6 x 2000mL (12L) capacity at 4,700 max rpm (7,187 x g max RCF) operation;
- 8 x 2000mL (16L) capacity at 3,900 max rpm (5,374 x g max RCF) operation
- Easy-to-use innovative 2000mL bucket and bottle with a wide mouth design requires no bridge and are resistant to 1000 cycles
- Thermo Scientific™ Eco-Spin™ windshield provides energy savings up to 60% when compared with non-windshielded rotors
- Compatible with Thermo Scientific™ Auto-Lid™ and Auto-ID™ enabling instant lid opening, storage and rotor identification that saves time and protects the integrity of your samples
- 2-year warranty
Upgrade to Thermo Scientific™ Dura-Coat™ buckets with nickel-coating to improve protection against corrosion due to moisture, chemicals or alkaline solutions.