Learn More
RETSCH 203.2 dia. x 25.4mmH Stainless Steel Test Sieve, ASTM Certificate, Micrometer Pore Sizes
Brand:  RETSCH 60.141.000250
 View more versions of this product
Product Code. 11984642
  • 114.00 € / Each
Estimated Shipment: 21-05-2025
to see stock.
Explore more special offers
Add to basket
Specifications
1Each
Test Sieve
Stainless Steel
25.4mm
203.2 mm
Stainless Steel
250 μm
ASTM E 11
Shaker AS 300
For separation, fractioning, particle size determination
Stainless Steel