Learn More
RETSCH 203.2 dia. x 25.4mmH Stainless Steel Test Sieve, ASTM Certificate, Micrometer Pore Sizes
114.00 € - 805.00 €
Specifications
Quantity | 1Each |
---|---|
Type | Test Sieve |
Certifications/Compliance | ASTM E 11 |
Material | Stainless Steel |
For Use With (Equipment) | Shaker AS 300 |
Product Code | Brand | Pore Size | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Pore Size | Price | Quantity & Availability | ||||||
11934642
|
RETSCH
60.141.000106 |
106 μm |
114.00 €
Each |
Estimated Shipment: 12-06-2025
Log in to see stock. |
||||||
11944642
|
RETSCH
60.141.000125 |
125 μm |
114.00 €
Each |
Estimated Shipment: 12-06-2025
Log in to see stock. |
||||||
11944632
|
RETSCH
60.141.000020 |
20 μm |
805.00 €
Each |
Estimated Shipment: 12-06-2025
Log in to see stock. |
||||||
11974642
|
RETSCH
60.141.000212 |
212 μm |
114.00 €
Each |
Estimated Shipment: 12-06-2025
Log in to see stock. |
||||||
11954642
|
RETSCH
60.141.000180 |
180 μm |
114.00 €
Each |
Estimated Shipment: 12-06-2025
Log in to see stock. |
||||||
11964642
|
RETSCH
60.141.000150 |
150 μm |
114.00 €
Each |
Estimated Shipment: 12-06-2025
Log in to see stock. |
||||||
11954632
|
RETSCH
60.141.000025 |
25 μm |
424.00 €
Each |
Estimated Shipment: 12-06-2025
Log in to see stock. |
||||||
11984642
|
RETSCH
60.141.000250 |
250 μm |
114.00 €
Each |
Estimated Shipment: 12-06-2025
Log in to see stock. |
||||||
11994642
|
RETSCH
60.141.000300 |
300 μm |
114.00 €
Each |
Estimated Shipment: 12-06-2025
Log in to see stock. |
||||||
11904652
|
RETSCH
60.141.000355 |
355 μm |
114.00 €
Each |
Estimated Shipment: 12-06-2025
Log in to see stock. |
||||||
11964632
|
RETSCH
60.141.000032 |
32 μm |
245.00 €
Each |
Estimated Shipment: 12-06-2025
Log in to see stock. |
||||||
11974632
|
RETSCH
60.141.000038 |
38 μm |
245.00 €
Each |
Estimated Shipment: 12-06-2025
Log in to see stock. |
||||||
11984632
|
RETSCH
60.141.000045 |
45 μm |
172.00 €
Each |
Estimated Shipment: 12-06-2025
Log in to see stock. |
||||||
11934652
|
RETSCH
60.141.000500 |
500 μm |
114.00 €
Each |
Estimated Shipment: 12-06-2025
Log in to see stock. |
||||||
11914652
|
RETSCH
60.141.000425 |
425 μm |
114.00 €
Each |
Estimated Shipment: 12-06-2025
Log in to see stock. |
||||||
11994632
|
RETSCH
60.141.000053 |
53 μm |
151.00 €
Each |
Estimated Shipment: 12-06-2025
Log in to see stock. |
||||||
11944652
|
RETSCH
60.141.000600 |
600 μm |
118.00 €
Each |
Estimated Shipment: 12-06-2025
Log in to see stock. |
||||||
11954652
|
RETSCH
60.141.000710 |
710 μm |
118.00 €
Each |
Estimated Shipment: 12-06-2025
Log in to see stock. |
||||||
11904642
|
RETSCH
60.141.000075 |
75 μm |
114.00 €
Each |
Estimated Shipment: 12-06-2025
Log in to see stock. |
||||||
11914642
|
RETSCH
60.141.000063 |
63 μm |
114.00 €
Each |
Estimated Shipment: 12-06-2025
Log in to see stock. |
||||||
11924642
|
RETSCH
60.141.000090 |
90 μm |
114.00 €
Each |
Estimated Shipment: 12-06-2025
Log in to see stock. |
||||||
11964652
|
RETSCH
60.141.000850 |
850 μm |
118.00 €
Each |
Estimated Shipment: 12-06-2025
Log in to see stock. |
||||||
Specifications
1Each | |
ASTM E 11 | |
Shaker AS 300 | |
For separation, fractioning, particle size determination | |
Stainless Steel |
Test Sieve | |
Stainless Steel | |
25.4mm | |
203.2 mm | |
Stainless Steel |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
RETSCH 203.2 dia. x 25.4mmH Stainless Steel Test Sieve, ASTM Certificate, Micrometer Pore Sizes