Learn More
RETSCH 203.2 dia. x 50.8mmH Stainless Steel Test Sieve, ISO Certificate, Micrometer Pore Sizes
Brand:  RETSCH 60.149.000630
 View more versions of this product
Product Code. 10298871
  • 121.00 € / Each
Estimated Shipment: 10-06-2025
to see stock.
Explore more special offers
Add to basket
Specifications
1Each
Test Sieve
Stainless Steel
50.8mm
203.2 mm
Stainless Steel
630 μm
ISO 3310/1
Shaker AS 300
For separation, fractioning, particle size determination
Stainless Steel