missing translation for 'onlineSavingsMsg'
Learn More
3-(4-Thiazolyl)-L-alanine, 95%, Thermo Scientific Chemicals Product Code.: 15460249

3-(4-Thiazolyl)-L-alanine, 95%, Thermo Scientific Chemicals

Product Code. 15460249
1 g
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 15460249

Brand: Thermo Scientific Alfa Aesar H63511.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Store at (-20°C). Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 119433-80-6
Molecular Formula C6H8N2O2S
Molecular Weight (g/mol) 172.202
MDL Number MFCD00079677
InChI Key WBZIGVCQRXJYQD-YFKPBYRVSA-N
Synonym 3-4-thiazolyl-l-alanine, s-2-amino-3-thiazol-4-yl propanoic acid, thiazolylalanine, 3-4-thiazoyl-l-alanine, unii-vtl1y3s44b, 2s-2-amino-3-1,3-thiazol-4-yl propanoic acid, vtl1y3s44b, s-3-4-thiazolyl alanine, l-thiazolylalanine, thiazolylalanine, l
PubChem CID 2761495
IUPAC Name (2S)-2-amino-3-(1,3-thiazol-4-yl)propanoic acid
SMILES C1=C(N=CS1)CC(C(=O)O)N

Specifications

Color Yellow
Solubility Information Slightly soluble in water.
Quantity 1 g
Formula Weight 172.2
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 3-(4-Thiazolyl)-L-alanine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberQ19H070 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH63511.03
Lot Number81301676 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberH63511.03
Lot NumberN11H027 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberH63511.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-(4-Thiazolyl)-L-alanine, 95%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.