missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Bromoimidazo[1,2-a]pyridine, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
10 g
Specifications
Specifications
Assay Percent Range | 97% |
Packaging | Amber glass bottle |
Quantity | 1 g |
Synonym | 3-bromoimidazo 1,2-a pyridine, 3-bromo-imidazo 1,2-a pyridine, 3-bromopyrimidazole, imidazo 1,2-a pyridine, 3-bromo, 3-bromoimidazolo 1,2-a pyridine, 3-bromoh-imidazo 1,2-a pyridine, pubchem17183, acmc-209kfj, buttpark 96\50-11, imidazo 1,2-a pyridine,3-bromo |
Formula Weight | 197.03 |
Percent Purity | 97% |
Chemical Name or Material | 3-Bromoimidazo[1, 2-a]pyridine |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 9
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
3-Bromoimidazo[1,2-a]pyridine, 97%, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction