missing translation for 'onlineSavingsMsg'
Learn More
3'-Deoxy-2',3'-didehydrothymidine, 98%, Thermo Scientific Chemicals Product Code.: 15288155

3'-Deoxy-2',3'-didehydrothymidine, 98%, Thermo Scientific Chemicals

Product Code. 15288155
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 15288155

Brand: Thermo Scientific Acros 454940010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3056-17-5
Molecular Formula C10H12N2O4
Molecular Weight (g/mol) 224.22
MDL Number MFCD00132921
InChI Key XNKLLVCARDGLGL-HGXVMFPFNA-N
Synonym stavudine, sanilvudine, zerit, 2',3'-didehydro-3'-deoxythymidine, estavudina, stavudinum, zerit xr, zerut xr, stavudinum inn-latin, 2',3'-anhydrothymidineShow MoreShow Less
PubChem CID 18283
ChEBI CHEBI:63581
IUPAC Name 1-[(2R,5S)-5-(hydroxymethyl)-2,5-dihydrofuran-2-yl]-5-methylpyrimidine-2,4-dione
SMILES CC1=CN([C@@H]2O[C@H](CO)C=C2)C(=O)NC1=O

Specifications

Melting Point 162.0°C to 169.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Merck Index 158929
Formula Weight 224.21
Percent Purity 98%
Chemical Name or Material 3'-Deoxy-2',3'-didehydrothymidine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0340738 Certificate TypeCertificate of Analysis Date18/01/2023 Catalog Number454940050,454940010
Lot NumberA0340738 Certificate TypeCertificate of Analysis Date15/12/2020 Catalog Number454940050,454940010
compliance-icons compliance-icons
Product Identifier
  • 3'-Deoxy-2',3'-didehydrothymidine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H341-Suspected of causing genetic defects.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3'-Deoxy-2',3'-didehydrothymidine, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.