missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ Disposable Protective Coveralls 4540+, Hooded
Coveralls are designed to help protect against hazardous dusts (Type 5) and certain light liquid splashes (Type 6).
236.00 € - 260.00 €
Specifications
Material | Polyethylene, Polyester/Polyethylene Laminate |
---|---|
Color | White |
Disposable | Single-use |
Waist Style | Elastic |
Ankle Style | Elastic |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
12652565
|
3M™
7100040087 |
Medium |
250.00 €
Pack of 20 |
Estimated Shipment: 07-03-2025
Log in to see stock. |
|||||
11903794
|
3M™
7100040416 |
Large |
250.00 €
Pack of 20 |
Estimated Shipment: 07-03-2025
Log in to see stock. |
|||||
11993784
|
3M™
7100040415 |
Small |
250.00 €
Pack of 20 |
Estimated Shipment: 09-04-2025
Log in to see stock. |
|||||
12662595
|
3M™
7100040086 |
X-Large |
250.00 €
Pack of 20 |
Estimated Shipment: 19-02-2025
Log in to see stock. |
|||||
12672595
|
3M™
7100036529 |
2X-Large |
260.00 €
Pack of 20 |
Estimated Shipment: 19-02-2025
Log in to see stock. |
|||||
11943794
|
3M™
7100041420 |
4X-Large | N/A | ||||||
11933794
|
3M™
7100041597 |
3X-Large |
250.00 €
Pack of 20 |
Estimated Shipment: 09-04-2025
Log in to see stock. |
|||||
Descrizione
- Premium dual material technology provides both protective performance and comfort
- Barrier against dry particles and limited liquid chemical splash (CE Type 5/6)
- Soft and light, super low-linting laminated microporous material
- Back panel made of breathable SMS material for improved air circulation and reduced heat stress
- Two-way zipper with fully sealable storm flap
- Three-panel hood design for enhanced compatability with complimentary PPE
- Knitted cuffs and elasticated waist and ankles for increased comfort and freedom of movement
- Anti-static treated
- Low linting
Specifica
Polyethylene, Polyester/Polyethylene Laminate | |
Single-use | |
Elastic | |
Serged, Bound |
White | |
Elastic | |
3-panel | |
Anti-static |
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto