missing translation for 'onlineSavingsMsg'
Learn More
4-Acetoxystyrene, 96%, stabilized with 200-300 ppm MEHQ, Thermo Scientific Chemicals Product Code.: 10294634

4-Acetoxystyrene, 96%, stabilized with 200-300 ppm MEHQ, Thermo Scientific Chemicals

Product Code. 10294634
5 mL, Glass bottle
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10294634

Brand: Thermo Scientific Acros 427000050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2628-16-2
Molecular Formula C10H10O2
Molecular Weight (g/mol) 162.19
MDL Number MFCD00075734
InChI Key JAMNSIXSLVPNLC-UHFFFAOYSA-N
Synonym 4-acetoxystyrene, 4-vinylphenyl acetate, 4-ethenylphenol acetate, p-acetoxystyrene, phenol, 4-ethenyl-, acetate, p-vinylphenol acetate, unii-7s21904ayn, phenol, 4-ethenyl-, 1-acetate, 4-ethenylphenyl acetate, acetic acid 4-vinylphenyl esterShow MoreShow Less
PubChem CID 75821
IUPAC Name (4-ethenylphenyl) acetate
SMILES CC(=O)OC1=CC=C(C=C1)C=C

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 7.0°C to 8.0°C
Color Colorless
Density 1.0600g/mL
Boiling Point 260.0°C
Flash Point 88°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Linear Formula CH3COOC6H4CH=CH2
Refractive Index 1.5375 to 1.5395 (20°C, 589nm)
Quantity 5 mL
Specific Gravity 1.06
Solubility Information Solubility in water: 0.1g/L (20°C)
Stabilizer stabilized with 200-300ppm MeHQ
Formula Weight 162.19
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 4-Acetoxystyrene, Stabilized with 200 to 300 ppm MEHQ
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0431451 Certificate TypeCertificate of Analysis Date05/01/2023 Catalog Number427000250,427000050
Lot NumberA0431451 Certificate TypeCertificate of Analysis Date29/06/2021 Catalog Number427000250,427000050
Lot NumberA0401390 Certificate TypeCertificate of Analysis Date06/11/2020 Catalog Number427000250,427000050
compliance-icons compliance-icons
Product Identifier
  • 4-Acetoxystyrene, stabilized
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H315-Causes skin irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Acetoxystyrene, 96%, stabilized with 200-300 ppm MEHQ, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.