Learn More
4'-Bromo-2'-hydroxyacetophenone, 95%, Thermo Scientific Chemicals
36.00 € - 453.00 €
Chemical Identifiers
CAS | 30186-18-6 |
---|---|
Molecular Formula | C8H7BrO2 |
Molecular Weight (g/mol) | 215.05 |
MDL Number | MFCD03428533 |
InChI Key | LQCMMXGKEGWUIM-UHFFFAOYSA-N |
Synonym | 1-4-bromo-2-hydroxyphenyl ethanone, 4'-bromo-2'-hydroxyacetophenone, 4-bromo-2-hydroxyacetophenone, 1-4-bromo-2-hydroxyphenyl ethan-1-one, acetophenone,2-hydroxy-4-bromo, ethanone, 1-4-bromo-2-hydroxyphenyl, 2-acetyl-5-bromophenol, 1-4-bromo-2-hydroxy-phenyl ethanone, 1-4-bromo-2-hydroxy-phenyl-ethanone |
PubChem CID | 10727403 |
IUPAC Name | 1-(4-bromo-2-hydroxyphenyl)ethanone |
SMILES | CC(=O)C1=C(O)C=C(Br)C=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15477638
|
Thermo Scientific Alfa Aesar
H61137.MD |
250 mg | N/A | ||||||
15457638
|
Thermo Scientific Alfa Aesar
H61137.03 |
1 g |
115.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15467638
|
Thermo Scientific Alfa Aesar
H61137.06 |
5 g |
453.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
4'-Bromo-2'-hydroxyacetophenone is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4′-Bromo-2′-hydroxyacetophenone is used as a pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Chemical Identifiers
30186-18-6 | |
215.05 | |
LQCMMXGKEGWUIM-UHFFFAOYSA-N | |
10727403 | |
CC(=O)C1=C(O)C=C(Br)C=C1 |
C8H7BrO2 | |
MFCD03428533 | |
1-4-bromo-2-hydroxyphenyl ethanone, 4'-bromo-2'-hydroxyacetophenone, 4-bromo-2-hydroxyacetophenone, 1-4-bromo-2-hydroxyphenyl ethan-1-one, acetophenone,2-hydroxy-4-bromo, ethanone, 1-4-bromo-2-hydroxyphenyl, 2-acetyl-5-bromophenol, 1-4-bromo-2-hydroxy-phenyl ethanone, 1-4-bromo-2-hydroxy-phenyl-ethanone | |
1-(4-bromo-2-hydroxyphenyl)ethanone |
Specifications
30186-18-6 | |
MFCD03428533 | |
Slightly soluble in water. | |
CC(=O)C1=C(O)C=C(Br)C=C1 | |
215.05 | |
215.04 | |
Solid |
C8H7BrO2 | |
1-4-bromo-2-hydroxyphenyl ethanone, 4'-bromo-2'-hydroxyacetophenone, 4-bromo-2-hydroxyacetophenone, 1-4-bromo-2-hydroxyphenyl ethan-1-one, acetophenone,2-hydroxy-4-bromo, ethanone, 1-4-bromo-2-hydroxyphenyl, 2-acetyl-5-bromophenol, 1-4-bromo-2-hydroxy-phenyl ethanone, 1-4-bromo-2-hydroxy-phenyl-ethanone | |
LQCMMXGKEGWUIM-UHFFFAOYSA-N | |
1-(4-bromo-2-hydroxyphenyl)ethanone | |
10727403 | |
95% | |
4'-Bromo-2'-hydroxyacetophenone |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only