missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Butylbenzene-1-sulfonyl chloride, 97%, Thermo Scientific™
45.10 € - 98.20 €
Chemical Identifiers
CAS | 54997-92-1 |
---|---|
Molecular Formula | C10H13ClO2S |
Molecular Weight (g/mol) | 232.72 |
MDL Number | MFCD00173908 |
InChI Key | OVFZELSNOHIDEF-UHFFFAOYSA-N |
Synonym | 4-butylbenzene-1-sulfonyl chloride, 4-n-butylbenzenesulfonyl chloride, 4-n-butyl benzenesulphonyl chloride, 4-butyl-benzenesulfonyl chloride, 4-n-butylphenylsulfonyl chloride, 4-n-butylbenzenesulphonyl chloride, benzenesulfonyl chloride, 4-butyl, 4-butylphenyl chlorosulfone, pubchem5484, 4-n-butylbenzenesulfonylchloride |
PubChem CID | 2737352 |
SMILES | CCCCC1=CC=C(C=C1)S(Cl)(=O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10447222
|
Thermo Scientific Maybridge
BTB10243EA |
10 g |
45.10 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10294974
|
Thermo Scientific Maybridge
BTB10243EB |
25 g |
98.20 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
54997-92-1 | |
232.72 | |
OVFZELSNOHIDEF-UHFFFAOYSA-N | |
2737352 |
C10H13ClO2S | |
MFCD00173908 | |
4-butylbenzene-1-sulfonyl chloride, 4-n-butylbenzenesulfonyl chloride, 4-n-butyl benzenesulphonyl chloride, 4-butyl-benzenesulfonyl chloride, 4-n-butylphenylsulfonyl chloride, 4-n-butylbenzenesulphonyl chloride, benzenesulfonyl chloride, 4-butyl, 4-butylphenyl chlorosulfone, pubchem5484, 4-n-butylbenzenesulfonylchloride | |
CCCCC1=CC=C(C=C1)S(Cl)(=O)=O |
Specifications
54997-92-1 | |
100.0 | |
Amber Glass Bottle | |
CH3(CH2)3C6H4SO2Cl | |
4-butylbenzene-1-sulfonyl chloride, 4-n-butylbenzenesulfonyl chloride, 4-n-butyl benzenesulphonyl chloride, 4-butyl-benzenesulfonyl chloride, 4-n-butylphenylsulfonyl chloride, 4-n-butylbenzenesulphonyl chloride, benzenesulfonyl chloride, 4-butyl, 4-butylphenyl chlorosulfone, pubchem5484, 4-n-butylbenzenesulfonylchloride | |
CCCCC1=CC=C(C=C1)S(Cl)(=O)=O | |
232.72 | |
232.73 | |
4-Butylbenzene-1-sulfonyl chloride |
96.5 | |
97% | |
C10H13ClO2S | |
MFCD00173908 | |
OVFZELSNOHIDEF-UHFFFAOYSA-N | |
4-butylbenzenesulfonyl chloride | |
2737352 | |
97% |
Safety and Handling
GHS P Statement Causes severe skin burns and eye damage. Contact with water liberates toxic gas. Reacts violently with water.
GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Immediately call a POISON CENTER or doctor/physician. IF IN EYES: Rinse cau
Danger