Learn More
4'-Chloroacetophenone, 98+%, Thermo Scientific Chemicals
49.85 € - 591.00 €
Chemical Identifiers
CAS | 99-91-2 |
---|---|
Molecular Formula | C8H7ClO |
Molecular Weight (g/mol) | 154.59 |
MDL Number | MFCD00000624 |
InChI Key | BUZYGTVTZYSBCU-UHFFFAOYSA-N |
Synonym | 4'-chloroacetophenone, 1-4-chlorophenyl ethanone, 4-chloroacetophenone, p-chloroacetophenone, ethanone, 1-4-chlorophenyl, acetophenone, 4'-chloro, 1-4-chlorophenyl ethan-1-one, 4-acetylchlorobenzene, p-chloracetophenone, p-acetylchlorobenzene |
PubChem CID | 7467 |
ChEBI | CHEBI:27538 |
IUPAC Name | 1-(4-chlorophenyl)ethanone |
SMILES | CC(=O)C1=CC=C(Cl)C=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11420053
|
Thermo Scientific Alfa Aesar
A12115.30 |
250 g |
49.85 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11410053
|
Thermo Scientific Alfa Aesar
A12115.0B |
1000 g |
153.77 €
1000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11430053
|
Thermo Scientific Alfa Aesar
A12115.0I |
5000 g | N/A | ||||||
Description
Chloroacetophenone is used as a tear gas for riot control and as a kind of chemical mace against small groups or individuals
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsChloroacetophenone is used as a tear gas for riot control and as a kind of chemical mace against small groups or individuals
Solubility
Soluble in water (0.11 mg/ml at 25°C), alcohol, and ether.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, strong bases.
Chemical Identifiers
99-91-2 | |
154.59 | |
BUZYGTVTZYSBCU-UHFFFAOYSA-N | |
7467 | |
1-(4-chlorophenyl)ethanone |
C8H7ClO | |
MFCD00000624 | |
4'-chloroacetophenone, 1-4-chlorophenyl ethanone, 4-chloroacetophenone, p-chloroacetophenone, ethanone, 1-4-chlorophenyl, acetophenone, 4'-chloro, 1-4-chlorophenyl ethan-1-one, 4-acetylchlorobenzene, p-chloracetophenone, p-acetylchlorobenzene | |
CHEBI:27538 | |
CC(=O)C1=CC=C(Cl)C=C1 |
Specifications
99-91-2 | |
1.19 | |
90°C (194°F) | |
1.555 | |
UN3416 | |
14,2116 | |
Soluble in water (0.11mg/ml at 25°C),alcohol,and ether. | |
CC(=O)C1=CC=C(Cl)C=C1 | |
154.59 | |
CHEBI:27538 | |
≥98% |
17°C to 20°C | |
230°C to 232°C | |
C8H7ClO | |
MFCD00000624 | |
386014 | |
4'-chloroacetophenone, 1-4-chlorophenyl ethanone, 4-chloroacetophenone, p-chloroacetophenone, ethanone, 1-4-chlorophenyl, acetophenone, 4'-chloro, 1-4-chlorophenyl ethan-1-one, 4-acetylchlorobenzene, p-chloracetophenone, p-acetylchlorobenzene | |
BUZYGTVTZYSBCU-UHFFFAOYSA-N | |
1-(4-chlorophenyl)ethanone | |
7467 | |
154.6 | |
4'-Chloroacetophenone |
Safety and Handling
GHS H Statement
H330-H227-H302-H315-H319-H335
Fatal if inhaled.
Combustible liquid.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P260-P264b-P270-P271-P280-P284-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
H302-H315-H318-H330-H335
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: CHLOROACETOPHENONE, LIQUID
missing translation for 'einecsNumber' : 202-800-7
missing translation for 'rtecsNumber' : KM5600000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- 4'-Chloroacetophenone
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
- MIXTURE LIST-Contains : 4'-Chloroacetophenone
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.