missing translation for 'onlineSavingsMsg'
Learn More
4-Methyl-1,2,3-thiadiazole-5-carbonyl chloride, Thermo Scientific™ Product Code.: 10130922

4-Methyl-1,2,3-thiadiazole-5-carbonyl chloride, Thermo Scientific™

Product Code. 10130922
1 g, Amber Glass Bottle
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy

Product Code. 10130922

Brand: Thermo Scientific Maybridge CC02302DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 59944-65-9
Molecular Formula C4H3ClN2OS
Molecular Weight (g/mol) 162.59
MDL Number MFCD00173831
InChI Key SDNDOCTUXWLDIX-UHFFFAOYSA-N
Synonym 4-methyl-1,2,3-thiadiazole-5-carbonyl chloride, 4-methyl-1,2,3-thiadiazole-5-carbonylchloride, 1,2,3-thiadiazole-5-carbonylchloride, 4-methyl, 4-methyl-1,2,3 thiadiazole-5-carbonyl chloride, 4-methyl-1,2,3-thiadiazole-5-carboxylic acid chloride, 4-methyl-5-thiadiazolecarbonyl chloride, 5-chlorocarbonyl-4-methyl-1,2,3-thiadiazole, 1,2,3-thiadiazole-5-carbonyl chloride, 4-methyl-9ciShow MoreShow Less
PubChem CID 2736982
SMILES CC1=C(SN=N1)C(Cl)=O

Specifications

Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 1 g
IUPAC Name 4-methylthiadiazole-5-carbonyl chloride
Formula Weight 162.6
Percent Purity 90%
Chemical Name or Material 4-Methyl-1, 2, 3-thiadiazole-5-carbonyl chloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Methyl-1,2,3-thiadiazole-5-carbonyl chloride, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.