missing translation for 'onlineSavingsMsg'
Learn More
4-(Methylsulfonyl)phenylhydrazine Hydrochloride, 95%, Thermo Scientific Chemicals Product Code.: 10230001

4-(Methylsulfonyl)phenylhydrazine Hydrochloride, 95%, Thermo Scientific Chemicals

Product Code. 10230001
2.5 g, Glass bottle
Click to view available options
Quantity:
2.5 g
10 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10230001

Brand: Thermo Scientific Acros 424960025

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17852-67-4
Molecular Formula C7H10N2O2S·HCl
Molecular Weight (g/mol) 222.69
MDL Number MFCD00216494
InChI Key QVCMFSJTNVQJFG-UHFFFAOYSA-N
Synonym 4-methylsulfonyl phenyl hydrazine hydrochloride, 4-methylsulfonyl phenylhydrazine hydrochloride, 4-methylsulphonyl phenylhydrazine hydrochloride, 4-methylsulphonylphenylhydrazine hydrochloride, 4-methanesulfonylphenyl hydrazine hydrochloride, 4-methylsulfonylphenylhydrazine hydrochloride, hydrazine, 4-methylsulfonyl phenyl-, monohydrochloride, 4-methylsulfonyl phenylhydrazine, chloride, 4-methylsulfonyl phenylhydrazine hydrochloric acid
PubChem CID 2735181
IUPAC Name (4-methylsulfonylphenyl)hydrazine;hydrochloride
SMILES CS(=O)(=O)C1=CC=C(C=C1)NN.Cl

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 202°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 94% min. (Argentometry)
Packaging Glass bottle
Linear Formula H2NHNC6H4SO2CH3·HCl
Quantity 2.5 g
Solubility Information Solubility in water: soluble
Formula Weight 222.69
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material 4-(Methylsulfonyl)phenylhydrazine hydrochloride
Show More Show Less
Product Identifier
  • 4-(Methylsulfonyl)phenylhydrazine hydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-(Methylsulfonyl)phenylhydrazine Hydrochloride, 95%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.