Learn More
RETSCH 400 dia. x 65 mmH Stainless Steel Test Sieve with Square Holes, ISO Certified
Brand: RETSCH 60.154.125000
Specifications
1Each | |
Test Sieve | |
Stainless Steel | |
For separation, fractioning, particle size determination | |
Stainless Steel |
125 mm | |
ISO 3310/2 | |
65mm | |
400 mm | |
Stainless Steel |
Product Suggestions
Customers who viewed this item also viewed.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.