Learn More
Brady™ Vinyl Tapes (B-595) for BMP71 Printer - Gold
Indoor/Outdoor vinyl grade conforms to irregular, smooth or rough surfaces
Brand:  Brady™ 142369
 View more versions of this product
Product Code. 11775633
Description
Description
  • Vinyl film with permanent adhesive
  • Adheres well to pipes, walls, equipment, windows - most clean, dry surfaces
  • Resistant to grease, oil and most industrial chemicals
  • Five year outdoor durability in temperatures from 82° to -40°C (180° to -40°F, can be applied as low as -18°C (0°F)
Specifications
Specifications
BS5609:1986 Section 2 and section 3
Permanent Acrylic
Gloss
Vinyl Tape
BMP™71 Printer
1 Ea.
0.102 mm
50 ft.
0.5 in.
BMP71 Label Printer Tapes
Gold
Vinyl (B-595)
Labeling pipes, walls, equipment, windows
1
-40°C to +82°C
15.24 m x 1.27 cm (50 ft. x 0.5 in.)
15.24 m
12.70 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Brady™ Vinyl Tapes (B-595) for BMP71 Printer - Gold > Gold; Size: 15.24m x 1.27cm (50 ft. x 0.5 in.)