Learn More
500 NTU Ratio Turbidity Standard, Reagecon
BIOPHARMA
Suspension of extremely accurate (± 1%) polymer microspheres with guaranteed shelf life stability even after opening the bottle.
Brand:  Reagecon CRSR-500-500
Product Code. 11526593
  • 375.00 € / 500mL
Explore more special offers
Description
Description
  • Turbidity Reference Standard, Turbidity Reference Material.
  • Designed and formulated to overcome the adverse safety, accuracy and stability issues experienced with other turbidity standards, primarily formazin-based standards.
  • Certified
  • NIST traceable
  • Non-toxic, non-carcinogenic
  • Highly accurate
  • Stable
  • EPA approved
Specifications
Specifications
Ratio Instruments
Accuracy ± 1% NTU
500 mL
Liquid format in HDPE Bottle GL45
500 NTU ± 1%
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
3 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberCRSR50024J1 Certificate TypeCertificate of Analysis Date02/04/2025 Catalog NumberCRSR-500-500
Lot NumberCRSR50023A1 Certificate TypeCertificate of Analysis Date08/05/2024 Catalog NumberCRSR-500-500
Lot NumberCRSR50021G1 Certificate TypeCertificate of Analysis Date07/10/2022 Catalog NumberCRSR-500-500
Special Offers
Special Offers
Product Content Correction
Product Title
500 NTU Ratio Turbidity Standard, Reagecon > 500 mL