missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spectrum™ Labs 0.1 - 0.5 kD Biotech CE Dialysis Kits
With tubing made from biologically inert and ultra-pure Biotech cellulose ester membrane
255.00 € - 310.00 €
Specifications
MWCO | 0.1 to 0.5 kDa |
---|---|
Content And Storage | Recommended storage in refrigeration (4-8°C). |
Includes:
- 1m Biotech CE Tubing in 0.05% sodium azide
- Two universal tubing closures
- Five opening picks
Product Code | Brand | Diameter (Metric) | Length (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Length (Metric) | Price | Quantity & Availability | ||||
15340662
|
Spectrum™
131060T |
20 mm | 3.1 mL/cm vol./length |
310.00 €
1m |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15330662
|
Spectrum™
131057T |
15 mm | 1.8 mL/cm vol./length |
300.00 €
1m |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15320662
|
Spectrum™
131048T |
6.4 mm | 0.32 mL/cm vol./length |
255.00 €
1m |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11570940
|
Spectrum™
131054T |
10 mm | 0.79 mL/cm vol./length |
278.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Ideal for small, critical dialysis applications that require the more concise selectivity and retention provided by Biotech Cellulose Ester (CE) Membrane than Standard Grade RC
- Hydrophilic, symmetric porositiy
- For small tubing demand applications or evaluating a specific dialysis membrane
- Includes 1m tubing with pair of tubing closures and 5 opening picks
Specifications
0.1 to 0.5 kDa | |
Recommended storage in refrigeration (4-8°C). |