missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spectrum™ Labs 0.1 - 0.5 kD Biotech CE Dialysis Kits
With tubing made from biologically inert and ultra-pure Biotech cellulose ester membrane
Brand: Spectrum™ 131048T
161.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Includes:
- 1m Biotech CE Tubing in 0.05% sodium azide
- Two universal tubing closures
- Five opening picks
Description
Ideal for small, critical dialysis applications that require the more concise selectivity and retention provided by Biotech Cellulose Ester (CE) Membrane than Standard Grade RC
- Hydrophilic, symmetric porositiy
- For small tubing demand applications or evaluating a specific dialysis membrane
- Includes 1m tubing with pair of tubing closures and 5 opening picks
Specifications
0.1 to 0.5 kDa | |
0.32 mL/cm vol./length | |
Recommended storage in refrigeration (4-8°C). |
6.4 mm | |
1 m Tubing/Kit |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 11
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spectrum™ Labs 0.1 - 0.5 kD Biotech CE Dialysis Kits > 10mm width; 6.4mm dia.; Vol./Length: 0.32mL/cm
Spot an opportunity for improvement?Share a Content Correction