Learn More
Spectrum™ Labs 0.1 - 0.5 kD Biotech CE Dialysis Kits
With tubing made from biologically inert and ultra-pure Biotech cellulose ester membrane
Brand: Spectrum™ 131057T
179.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Includes:
- 1m Biotech CE Tubing in 0.05% sodium azide
- Two universal tubing closures
- Five opening picks
Description
Ideal for small, critical dialysis applications that require the more concise selectivity and retention provided by Biotech Cellulose Ester (CE) Membrane than Standard Grade RC
- Hydrophilic, symmetric porositiy
- For small tubing demand applications or evaluating a specific dialysis membrane
- Includes 1m tubing with pair of tubing closures and 5 opening picks
Specifications
0.1 to 0.5 kDa | |
1.8 mL/cm vol./length | |
Recommended storage in refrigeration (4-8°C). |
15 mm | |
1 m Tubing/Kit |