missing translation for 'onlineSavingsMsg'
Learn More
6-Oxa-1-azaspiro[3.3]heptane hemioxalate, 95%, Thermo Scientific Chemicals Product Code.: 15404108

6-Oxa-1-azaspiro[3.3]heptane hemioxalate, 95%, Thermo Scientific Chemicals

Product Code. 15404108
500 mg
Click to view available options
Quantity:
250 mg
500 mg
This item is not returnable. View return policy

Product Code. 15404108

Brand: Thermo Scientific Alfa Aesar H57658.ME

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Notes
Incompatible with strong oxidizing agents, strong acids and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1380571-72-1
Molecular Formula C12H20N2O6
Molecular Weight (g/mol) 288.3
MDL Number MFCD22373634
InChI Key UMKDEMSXCWMHRS-UHFFFAOYSA-N
Synonym 6-oxa-1-azaspiro 3.3 heptane oxalate 2:1, 6-oxa-1-azaspiro 3.3 heptane hemioxalate, 6-oxa-1-azaspiro 3.3 heptane oxalate 2, bis 6-oxa-1-azaspiro 3.3 heptane oxalate, 6-oxa-1-azaspiro 3.3 heptane,ethanedioate 2:1, oxalic acid-6-oxa-1-azaspiro 3.3 heptane 1/2, bis 6-oxa-1-azaspiro 3.3 heptane ; oxalic acidShow MoreShow Less
PubChem CID 71299818
IUPAC Name 6-oxa-1-azaspiro[3.3]heptane;oxalic acid
SMILES C1CNC12COC2.C1CNC12COC2.C(=O)(C(=O)O)O

Specifications

Quantity 500 mg
Formula Weight 144.15
Percent Purity 95%
Chemical Name or Material 6-Oxa-1-azaspiro[3.3]heptane hemioxalate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberU21A007 Certificate TypeCertificate of Analysis Date21/01/2025 Catalog NumberH57658.MD,H57658.ME
Lot NumberX11H001 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH57658.MD,H57658.ME
Lot NumberB13Y056 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH57658.MD,H57658.ME
Lot NumberQ10A010 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH57658.MD,H57658.ME
Lot NumberU21A007 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH57658.MD,H57658.ME

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
6-Oxa-1-azaspiro[3.3]heptane hemioxalate, 95%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.