Learn More
Testo™ 606-2 - Moisture Meter
Measures the moisture content in wood, building materials, and the surrounding environment. This allows you to check storage and drying conditions quickly and easily.
Brand: Testo™ 0560 6062
This item is not returnable.
View return policy
Description
- Measures the moisture content of different types of wood and building materials, and is also able to measure relative humidity and air temperature, and calculate the dew point and wet bulb temperature
- Ram electrodes guarantee precise moisture measurements in wood / building materials
- Armed with such valuable information, you can now easily determine whether the storage conditions are ideal, or whether they need to be adjusted
- The characteristic curves allow the moisture content to be displayed directly as a weight percentage based on the dry mass (dry weight). The following characteristic curves are stored in the meter:
- for precise moisture measurements in beech, spruce, larch, oak, pine, maple
- for detecting moisture and wet places in building materials like cement screed, concrete, plaster, anhydrite screed, cement mortar, limestone mortar, and brickwork
- Designed to make work more efficient and life easier - It has a variety of practical features including:
- a hold function, which freezes the last reading
- a backlit display for use in all lighting conditions
- a slip-on protective cap, and a belt bag, which ensure that your moisture meter is stored safely
- a hand strap that helps prevent your meter from being accidentally dropped
Specifications
±1% | |
119 mm | |
46 mm |
25 mm | |
0.1°C |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Product Title
Testo™ 606-2 - Moisture Meter > Resolution: 0.1°C
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu