Learn More
Digi-Sense™ Precalibrated Pocket Size Moisture Meter
Rugged pocket sized meter with one-handed operation
Brand: Digi-Sense™ 20250-35
Includes: Four LR44 batteries and NIST-traceable calibration report with data supplied by InnoCal.
Description
- Reliable measurements guaranteed
- Precalibrated to NIST-traceable standards
- Measure the moisture level in wood and hardened materials
- Low battery indication
Services
2 Years
Specifications
Wood: ±2%; Material: ±0.2%; Temp: ±1.8° F (1°C) | |
Four LR44 batteries and NIST-traceable calibration report with data supplied by InnoCal. | |
4 cm |
2 cm | |
9 cm | |
Moisture Meter |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.