Learn More
Jenway™ 72 Series Genova Bio Spectrophotometer
The Genova Bio is a simple, low cost, easy to use UV/visible spectrophotometer; dedicated for life science applications. The fast, responsive colour touchscreen and pre-programmed methods for the measurement of nucleic acid concentration and purity, protein assays and cell density, makes this spectrophotometer the ideal addition to any laboratory.
Brand: Jenway™ 83056-04
3641.62 EUR valid until 2025-12-31
Use promo code "24356" to get your promotional price.
Description
- Life Science Spectrophotometer
- Pre-programmed for DNA, RNA and protein analysis
- Compatible with ultra-micro, semi-micro, micro and macro cuvettes
- Scanning diode array technology
- Color touchscreen navigation
- Small footprint and lightweight (< 3kg)
- Fast scan speed
- English, French, German, Italian and Spanish language options
- Multiple USB ports for data storage and printer connectivity
- 2-year warranty including xenon lamp
Specifications
±0.01A at 1.0A and 546nm | |
15 mm | |
Photodiode Array | |
100/240 V | |
50/60 Hz | |
±0.002A at 0.1A and ±0.02A at 2.0A and 546nm | |
±2 nm | |
At 340 nm: < 1%T according to ANSI/ASTM | |
±2 nm | |
2.8 kg |
−0.300 to +2.500 A | |
±2500 | |
3 nm | |
198 to 800 nm | |
Color touch screen | |
Photodiode Array | |
Optional | |
2 years including Xenon lamp | |
±2 nm |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.