missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ γ-Globulin, Human Plasma, 1g
Each donor unit used in the preparation of the product was tested and found negative for Anti-HIV 1/2, HBsAg, Anti-HCV and HIV-1 Ag by FDA approved methods or CE approved methods.
Brand: MP Biomedicals™ 08823101
114.00 EUR valid until 2025-05-31
Use promo code "25977" to get your promotional price.
This item is not returnable.
View return policy
Description

Specifications
White | |
1 g | |
Powder | |
>96% |
6.44 | |
2°C to 8°C | |
Used as an immunological challenge in studies of mechanisms of immunotolerance | |
Human Plasma gamma Globulin |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
MP Biomedicals™ γ-Globulin, Human Plasma, 1g > Human Plasma, 1g
Spot an opportunity for improvement?Share a Content Correction