Learn More
MP Biomedicals™ γ-Globulin, Bovine Purified
Suitable for use as a carrier protein in secondary antibody systems, e.g., radioimmunoassay. Purified proteins are recommended as ELISA coating antigens and as controls.
Brand:  MP Biomedicals™ 0855847
Product Code. 17592936
  • 502.00 € / 100mg
Estimated Shipment: 29-05-2025
to see stock.
Explore more special offers
Add to basket
Specifications
20 mg/mL
2°C to 8°C
Immunoassay
100 mg
Lyophilized
Purified Bovine gamma Globulin
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
MP Biomedicals™ γ-Globulin, Bovine Purified > Bovine Purified, 100mg