missing translation for 'onlineSavingsMsg'
Learn More
A 769662, Tocris Bioscience™ Product Code.: 18034392

A 769662, Tocris Bioscience™

Product Code. 18034392
10 mg
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy

Product Code. 18034392

Brand: Tocris Bioscience 3336/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent AMPK activator

Potent, reversible AMP-activated protein kinase (AMPK) activator (EC50 = 0.8 μM) that displays selectivity towards β1 subunit-containing heterotrimers. Inhibits fatty acid synthesis (IC50 = 3.2 μM) and decreases plasma glucose and triglyceride levels in vivo. Also inhibits proliferation of mesenchymal stem cells, and impedes reprogramming of mouse embryonic fibroblasts to iPSCs. Activates autophagy.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 844499-71-4
Molecular Formula C20H12N2O3S
Molecular Weight (g/mol) 360.387
InChI Key CTESJDQKVOEUOY-UHFFFAOYSA-N
Synonym 4-hydroxy-3-2'-hydroxy-1,1'-biphenyl-4-yl-6-oxo-6,7-dihydrothieno 2,3-b pyridine-5-carbonitrile, unii-p68477cd2c, 6,7-dihydro-4-hydroxy-3-2'-hydroxy 1,1'-biphenyl-4-yl-6-oxo-thieno 2,3-b pyridine-5-carbonitrile, 4-hydroxy-3-2'-hydroxybiphenyl-4-yl-6-oxo-6,7-dihydrothieno 2,3-b pyridine-5-carbonitrile, 4-hydroxy-3-4-2-hydroxyphenyl phenyl-6-oxo-7h-thieno 2,3-b pyridine-5-carbonitrile, thieno 2,3-b pyridine-5-carbonitrile, 6,7-dihydro-4-hydroxy-3-2'-hydroxy 1,1'-biphenyl-4-yl-6-oxo, 4cff, 4-hydroxy-3-2'-hydroxy-1,1'-biphenyl-4-yl-6-oxo-7h-thieno 2,3-b pyridine-5-carbonitrileShow MoreShow Less
PubChem CID 54708532
IUPAC Name 4-hydroxy-3-[4-(2-hydroxyphenyl)phenyl]-6-oxo-7H-thieno[2,3-b]pyridine-5-carbonitrile
SMILES C1=CC=C(C(=C1)C2=CC=C(C=C2)C3=CSC4=C3C(=C(C(=O)N4)C#N)O)O

Specifications

Quantity 10 mg
Formula Weight 360.39
Percent Purity >98%
Chemical Name or Material A 769662
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
A 769662, Tocris Bioscience™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.