Learn More
Fisher Scientific™ Accu500 Water System
Exceeds ASTM, CLSI, CAP, and ISO Type I water standards. Fisher Scientific™ Accu500 Water System is a versatile purification system that can be used in a number of ways.
Brand: Fisher Scientific™ RD0P0100EK
This item is not returnable.
View return policy
Description
- The fully automated control system contains pre-rinse, a flush feature preventing RO membrane fouling, RO membrane and cartridge life monitoring as well as many other programs for convenient operation
- The pre-rinsing program starts automatically once a new RO membrane is installed so there is no need for user to set additional cleaning procedures
- Two sets of dual-column purification cartridges work sequentially to ensure thorough removal of ions and organics in water
- Automatic hourly system recirculation prevents bacterial growth
- Resistivity is measured by a highly sensitive resistivity sensor
- Electrical and water compartments are completely segregated, thus safer to operate
- User can set water dispensing time for easy water collection
- Both RO and ultrapure water can be dispensed from the water system
- The system can be fitted with a 30L or 60L PE tank with a liquid level sensor and a tank vent filter with CO2 scavenger to protect purified water from atmospheric degradation
- A 0.2μm filter or ultrafiltration filter (optional) removes final contaminants
- Independent ultrapure water dispenser
- The remote dispenser has a built-in high precision resistivity sensor with cell constant of 0.01cm-1 to measure the quality of ultrapure water just prior to the dispensing point
- Volumetric dispensing and calibration
- The handle is 360 degree-rotatable and detachable from its support for effortless and instant dispensing with maximum flexibility
Applications:
Ultrapure and RO water can be used in many areas. Some typical applications include:
Ultrapure water
- HPLC mobile phase preparation
- Preparation of reagent blank solutions
- As sample diluent for GC, HPLC, AA, ICP-MS and other analytical techniques
- Preparation of buffers and culture media for mammalian cell culture
- Preparation of molecular biology reagents, etc.
RO water
- Glassware cleaning
- Washing machine for glassware
- Waterbath water
- Autoclaving
Specifications
<20μS/cm | |
Tap Water | |
Accu500 Ultrapure Water System | |
Accu500 | |
1.5 L/min. | |
5°C to 45°C | |
48 cm |
<0.001 EU/mL with a terminal ultrafiltration cartridge | |
18.2 Mohm.cm | |
1 to 6 bar | |
<1/mL with 0.2μm final filter or terminal ultrafiltration cartridge | |
10 L/hr. | |
30 cm | |
51 cm |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Product Title
Fisher Scientific™ Accu500 Water System >
Spot an opportunity for improvement?Share a Content Correction